Orders LinkContactUsLinkComponent
Inverted-Microscopes.jpg
Leica Compound Inverted Microscopes
Inverted microscopes from Leica Microsystems offer the flexibility and power you need to advance your research and enable publication quality documentation.
Request a call
Leica Compound Inverted Microscopes | Avantor
Leica inverted microscopes offer you high performance to help you advance your research.
The Leica DMi1 inverted microscope supports your specific work routines in your cell culture lab. Its operation is so intuitive, its handling so comfortable that you can fully concentrate on your work. Choose the functions you need, and, if necessary, you can easily add a variety of accessories that are important for your work. With high performance optics, ergonomic design, and 5 W LED illumination, the Leica DM IL LED is ideal for cell culture, micromanipulation, documentation of immunostained specimens, and routine live cell examinations.
The Leica DM IL LED features a comprehensive set of contrast methods to monitor your specimen the way you need. High quality phase contrast, excellent modulation contrast and brilliant fluorescence are just one fingertip away. Robust stability, plenty of space to work with tools, long working distances to accommodate large culture flasks and a stable illumination without heat make work at the microscope easy and convenient.
For special diagnostics requirements, this microscope is certified for in-vitro diagnostics (IVD) including in-vitro fertilization (IVF)
dmi_260.jpg
DMi1
Just right for your live cell lab – smart choice for quick checks. The well designed and quality control elements enable you to work in comfort.
More info
dmi-led-260.jpg
DM IL LED
The Leica DM IL LED inverted routine microscope features a comprehensive set of contrast methods to monitor your specimen the way you need.
More info
mateo-260.jpg
Mateo TL
A digital microscope for routine cell culture with an intuitive set-up and use, making the process of checking cell culture growth quick and effortless for every member in your lab.
More info