Orders LinkContactUsLinkComponent
DehydratedCultureHeader.jpg
Criterion™ Dehydrated Culture Media by Hardy Diagnostics
CRITERION™: the logical choice for dehydrated culture media in your laboratory. | Avantor
Hardy Diagnostics’ dehydrated culture media, CRITERION™, is formulated to meet or exceed the highest quality standards. Choose from 250 standard formulas or request custom blending to your specifications. The ergonomic packaging has built in handles for an improved user experience, as well as made from opaque materials to diminish light penetration. Criterion™ jars and buckets are designed to be stackable to help save room during storage.
An induction seal, as well as a built-in cushion seal and desiccant pack, ensures that the media stays dry and does not clump. Jars and buckets are built with wide mouth lids allowing easy access to scoop the media during measuring; no more shaking the bottle to dispense the media!
The innovative packaging designs and overall reliability make CRITERION™ the logical choice for culture media in your laboratory or classroom.