Orders LinkContactUsLinkComponent
DWK Pipettes | Avantor
Pipettes
Our pipette line includes many application-specific varieties: bacteriological, serological, and measuring pipettes to name a few. In addition, we offer different characteristics to suit specific purposes including glass, plastic, and electronic versions plus a variety of sizes.
Pasteur-Pipettes400-495.jpg
Choose from pipettes guaranteed to accurately transfer or measure liquid samples.
Volumetric-Pipettes400-495.jpg
DWK volumetric Pipettes are manufactured according to both ASTM E543, and E969, are NIST traceable and will provide years of accurate service.
Bacteriological-Pipettes400-495.jpg
Glass Pipettes that are designed specifically for testing milk and milk products for bacterial contamination
Serological-Pipettes400-495.jpg
Available in sterile and non-sterile, disposable serological pipettes are manufactured from 51-expansion borosilicate glass.
Automatic-Manual-Pipettes400-495-.jpg
DWK's complete line of pipettes offers your lab a single source for superior quality pipettes