Orders LinkContactUsLinkComponent
Contec-Trolley08-KR-feature.jpg
Cleanroom Mopping Systems
Contec has developed the most complete line of mopping products, including wall and ceiling mops, buckets and tools for critical and cleanroom environments.
Whether disinfecting or cleaning floors, walls, ceilings, or inside bio-safety cabinets and isolators, Contec has a cleanroom mopping system or cleaning tool that meets the demanding requirements of today’s high-tech industries. Many of our cleanroom mops are available with sterile validation for use in aseptic environments.
Featured products
subpage3_VertiKlean_MAX2_155_155.jpg
This durable, lightweight mop head features a single layer of hydrophilic PU foam laminated to polyester microfibre fabric. The ribbed microfibre material smoothly glides over floors and textured surfaces providing superior contamination removal.
subpage3_QuickTaskFlatMop2_155_155.jpg
Contec’s QuickTask™ range consists of a variety of flat mop heads that are easily installed onto a hinged mop head frame and handle assembly. Each mop head has been specifically chosen to be compatible with a wide range of chemicals and disinfectants.
subpage3_MopSaturationSystem_155_155.jpg
Designed to simplify the cleanroom mopping process, the Mop Head Saturation System allows mop heads to be presaturated prior to being passed into the controlled environment. Small and compact, the stainless steel trolley is ideal for small cleanrooms.
subpage3_CompactBucketSystem_155_155.jpg
Contec’s autoclavable Compact Bucket System is an all-in-one solution to make cleaning and disinfecting easier and more efficient. The stainless steel trolley is available with either two or three autoclavable polypropylene buckets.
Videos
Whitepapers and Articles
Literature