ContactUsLinkComponent
Contect_hero955x555.jpg
So much more than a clean surface...
Contec Inc is an ISO 9001:2015 registered manufacturer of contamination control products for critical cleaning and disinfection in controlled environments worldwide. 
Contec’s extensive cleanroom product range of presaturated and dry wipes, mops, disinfectants and swabs; both sterile and non sterile is ideal for the many cleaning and disinfecting tasks in critical environments.
Contec cleanroom products are used in a variety of industries including pharmaceutical, medical device, biopharma, microelectronics, optics, semiconductor and electronics.
alcohols-and-solutions-240-150.jpg
A wide range of disinfectants and solutions for critical and sterile environments. From hydrogen peroxide solutions and IPA, to fast-acting validated sporicidal disinfectants. Contec can also offer a sterile neutral detergent solution.
wiipe_thumbnail_240_150.jpg
Sterile and non-sterile, dry and presaturated, knitted, nonwoven wipes and microfibre wipes, as well as a range of swabs of different lengths, head sizes, and materials.
mopping_thumbnail_240_150.jpg
A broad range of wall and ceiling mops, buckets, trolleys and tools for critical and cleanroom environments, whether disinfecting or cleaning floors, walls, ceilings, or inside bio-safety cabinets and isolators.
Our Featured Products
Refibe_Beach_Image_155_155.jpg
The 1st sustainable cleanroom polyester wipe made from recycled post-consumer plastic bottles. ReFIBE™ are 100% knitted polyester wipes with sealed edges, with each case stopping up to 840 bottles from going to landfill.
Opening_PS_pouch_155_155.jpg
Presaturated wipes offer many benefits over bottled alcohol and dry wipes. They reduce solvent use, increase convenience and provide exceptional solvent control. Contec's 70% IPA presat wipes are all authorised to the EU Biocidal Products Regulation.
ProChlor_spraying_155_155.jpg
Contec ProChlor is a revolutionary sporicide achieving a 100% kill against spores in under 1 min. A blend of hypochlorous acid in purified water, Contec ProChlor is provided ready-to-use and is efficacious against bacteria, fungi, viruses and spores.
EasyReach_in_use_155_155.jpg
Designed for cleaning isolators, laminar air flow hoods, glove boxes and other mini environments, Contec’s lightweight, stainless steel, EasyReach™ Cleaning Tool is easy to use and improves access to hard to reach areas.
Videos
Literature
Webinars
Whitepapers and Articles
Find the product that’s right for your facility