Orders LinkContactUsLinkComponent
Biotix Plates | Avantor
Biotix Plates
Biotix Assay and Deep Well Plates are designed for high throughput screening; engineered with raised alpha/numeric grids for clear well identification, sample tracking and plate orientation. All plates are manufactured with non-treated, medical-grade virgin polypropylene making them biologically inert with great temperature and chemical resistance.
Assay-Plates-400x495.jpg
Available in both U-well and V-well formats with wide wells making them ideal for a wide range of applications including combinatorial chemistry, fluorescence/luminescence assays, and compound libraries.
View our different Assay plate options here
Deep-Well-Plates-400x495.jpg
Deep Well Plates
Biotix deep well plates are engineered to be stackable with a SBS footprint for use with automation platforms. Great for use in ELISA assays, colorimetric assays and a wide range of other functions in cell culture and storage.
Click here to see our deep well plate options