ContactUsLinkComponent
Product Differentiation Table
FBS Products
Australia
Origin FBS
Ultimate
Grade FBS
Premium
Grade FBS
Select Grade FBSUltra Low
IgG FBS
Country of Origin100% US Origin XXX
Non-US OriginXX
Product FeaturesSingle-Use Filtration TechnologyX
Cold Temperature ManufacturedXXXX
Triple 0.1μm Sterile FilteredXXXXX
Whole-lot Homogenous TechnologyXXXXX
Independently Tested by Third-Party LaboratoriesXXXXX
Specifications
ISIA Traceability Certified SeraXXXX
Endotoxin**≤ 5≤ 5≤ 5≤ 10≤ 5
Hemoglobin**≤ 25≤ 25≤ 25≤ 25≤ 25
9CFR Virus Testing, Sterility, MycoplasmaXXXXX
Biochemical AssayXXXXX
Electrophoretic ProfileXXXXX
Additional Virus Testing by qPCR TechnologyX
*International Serum Industry Association (serumindustry.org)
**The care and attention devoted to the manufacture of our sera generates final test results that are substantially lower than set specifications.
See lot-specific Certificates of Analysis for actual test results.
ISIA255.jpg
ISIA Certified Traceability
The International Serum Industry Association (ISIA) has established industry standards to aid sera users in substantiating the traceability of serum products. The ISIA has implemented a rigid program of independent audits to verify compliance with traceability standards. ISIA Traceability Certification is awarded to those that demonstrate full compliance with ISIA guidelines and are the subject of a successful audit. Further, the ISIA has established strict guidelines for product quality testing and reporting on documents like Certificates of Analysis (CoA). Sera users should source exclusively from ISIA-certified companies to ensure traceability.
Oritain255.jpg
Oritain Partnership
Avantor is part of Oritain database. The Oritain verification of product origin provides enhanced traceability and trust for Avantor® Seradigm Fetal Bovine Serum (FBS). Oritain's independent, scientific sampling and analysis provides added confidence for customers using Seradigm FBS. By partnering with Oritain, we are able to verify the “fingerprint” of FBS origin exclusive to the source material country of origin. Confirmation by individual lot can be found on the product Certificates of Analysis (COA).