ContactUsLinkComponent
detector_parts.png
Detector Parts & Accessories
Find the right supplies for your GC detector. Agilent has a full list of parts for electron capture detectors (ECDs), flame ionization detectors (FIDs), flame photometric detectors (FPDs), thermal conductivity detectors (TCDs), nitrogen chemiluminescence detectors (NCDs), nitrogen phosphorus detectors (NPDs), and sulfur chemiluminescence detectors (SCDs).
Product Selection
detector_parts.png
The Agilent micro ECD is the most sensitive on the market, with a detection zone volume 10 times smaller than any other ECD.
FID.png
Flame ionization detection is the most commonly used GC detection method due to it's superior ability to measure hydrocarbons.
Flame_Photometric.png
The FPD uses three gases: air and hydrogen to support the flame, and nitrogen makeup for capillary columns.
Burner_Accessory.png
A nitrogen-specific detector that produces a linear and equimolar response to nitrogen compounds based on a chemiluminescent reaction of NO with ozone
NPD.png
NPD for GC are specific to nitrogen- or phosphorus-containing compounds and are great for environmental and forensic applications.
SulfurChemilumine.png
Sulfur Chemiluminescence Detector is the most sensitive and selective chromatographic sulfur detector available for the analysis of sulfur compounds.