ContactUsLinkComponent
Capillary_Tubing.png
Capillary Tubs and Accessories
Agilent provides all the necessary gas chromatography accessories and tooling for you to perform maintenance on your capillary columns. Our extensive portfolio includes column hangers and baskets, capillary rinse kits, installation kits, and preswaging tools.
Product Selection
Cap_Column.png
The capillary column installation kit provides everything needed to easily install a GC capillary column.
ColRinse.png
This kit makes solvent rinsing easy! Solvent rinsing removes many contaminants that accumulate in columns.
Capillary_Tubing.png
Agilent offers a range of column baskets and hangers
PreSwagingTools.png
Optimize GC performance and reproducibility by ensuring a proper length of column into the inlet, time after time.
FusedSilica.png
Fused Silica Tubing Cutters
Tool and ceramic wafer that allow you to make clean and easy cuts in fused silica, glass, and aluminum-clad capillary columns in GC systems.