ContactUsLinkComponent
chemicals-1920x760.jpg
VWR Chemicals
Avantor® offers a wide range of laboratory and industrial chemicals under our leading brands, which are recognized for high quality, detailed specifications, safe packaging, excellent service and outstanding technical support.
Our portfolio features everything from basic technical reagents to high-purity chemicals for trace analysis, plus extensive specialty ranges for applications like microbiology, cell diagnostics and life sciences.
Environmental testing
ph-strips-540x390.jpg
pH test strips provide precise and rapid pH determination, with up to four color blocks for each pH value.
photometric-tests-540x390.jpg
Powder pack reagents from VWR® Chemicals are designed for use with laboratory and portable photometers with 16-mm cell capability. No calibration data is necessary. The pillows, individually sealed in aluminum foil packs, are unaffected by ambient conditions and contain pre-measured powdered reagents for accuracy, convenience and high solubility.
33938798-540x390.jpg
Our VWR® Chemicals portfolio features a wide range of ISO 17034-certified reference materials for methods such as COD, TOC, turbidity measurements and photometry.
Chromatography
avantor_ace_hichrom-540x390.jpg
Avantor® ACE® columns incorporate the latest developments in LC stationary phase design to provide chromatographers with more choices for alternative selectivity, without compromising stability or robustness.
30881599.jpg
GC columns are suitable for analyzing a wide range of substances, such as solvents, alcohols, volatiles, pesticides, herbicides, insecticides and other materials.
VWR_BDH_Chemicals_540x390.jpg
HPLC sorbents are used primarily for low-pressure preparative chromatography. Mesh specifications ensure the product does not contain an excess of fines, which could affect flow rate and separation performance.
IC_standards_main.jpg
Avantor® offers a wide range of standards (54 elements at 1000 mg/L) for food and beverage, environmental, pharmaceutical and other chromatography processes.
23493492.jpg
Our wide range of secondary GC reference standards are available for various chromatographic and analytical applications.
Instrumental analysis
NMR-range-N_3-540x390.jpg
Avantor® offers a complete range of deuterated solvents with high purity (>99.9%) and varying degrees of deuteration, available in a variety of pack sizes.
36149988-540x390.jpg
Choose from our large portfolio of Karl Fischer volumetric, coulometric and standard reagents adapted to all titrators for water determination.
NORMATOM-ACIDS-N-540x390.jpg
A complete range of high-purity acids with impurities in ppb and ppt levels for sample preparation in metal trace analysis.
23395659-540x390.jpg
If you are searching for a specific volumetric solution, Avantor® can help you find the perfect reagent for your titrations.
Sodium-VWR-AAS-+-Certificat-N-540x390.jpg
Our AAS standards (1000 mg/L) are available in 100- and 500-ml pack sizes, traceable to NIST, ISO 17034 certified and delivered with a complete CoA.
Aluminium-ICP-standard-100-ml-540x390.jpg
Our comprehensive range of single- and multi-element solutions (available in 10, 1000, and 10 000 mg/L in 100- and 500-ml pack sizes) is traceable to NIST, ISO 17034 certified, and delivered with a complete CoA.
VWR-AVS-Titrinorm-satchets-540x390.jpg
Proper calibration protects your research by preventing incorrect results. Find our extensive range of pH buffers and conductivity solutions available in many different formats.
AT-density_9-540x390.jpg
See our wide range of certified reference materials for measuring physical properties such as viscosity, density, refractive index, osmolality and others.
Solvents
28506744-540x390.jpg
We provide a complete range of solvents for various analytical and R&D applications. Each has guaranteed specifications that conform to ACS and Reag. Ph. Eur. standards.
Product040-Toluene-RECTAPUR-5L-540x390.jpg
Select from a wide range of solvents with limited specifications for general laboratory use.
UHPLC_group4-540x390.jpg
A superior-quality mobile phase with high UV transmittance, low residue on evaporation and batch-to-batch reproducibility is essential for avoiding unwanted ghost peaks. Our high-purity solvents deliver the performance you need.
23395528-540x390.jpg
Our high-purity LC-MS solvents and reagents have low ionic background levels and low alkaline metal contents, making them suitable for UHPLC and LC-MS chromatography.
Solvents-for-preparative-8-540x390.jpg
Specially tailored for the requirements of preparative HPLC separations, these solvents have very low evaporation residue levels (<1 mg/L), low water contents and high purity.
hplc-solvents-540x390.jpg
Our complete range of HPLC solvents in stainless steel returnable barrels improves safety, limits potential contamination and eliminates waste.
28776-540x390.jpg
These high-purity solvents are designed for spectroscopy applications that demand maximum spectral purity.
NMR-range-N_3-540x390.jpg
Avantor® offers a complete range of deuterated solvents with high purity (>99.9%) and varying degrees of deuteration, available in a variety of pack sizes.
pestinorm_supratrace_solvents_main-540x390.jpg
For gas chromatography, particularly pesticide analysis, we offer a wide range of solvents with extremely low levels of halogenated and other organic compounds to prevent sample contamination.
ANHYDROUS-SOLVENT-N-540x390.jpg
Discover our complete portfolio of anhydrous solvents with low water contents (<0.005%), available in 250- and 500-ml septum-sealed glass bottles.
Inorganic reagents
23395524-540x390.jpg
We offer a comprehensive range of acids suitable for many analytical and R&D applications, with guaranteed specifications that conform to ACS and Reag. Ph. Eur. standards.
34454418-540x390.jpg
Learn about our acids for general laboratory use.
34454418-540x390.jpg
This wide range of acids with limited specifications is for general laboratory use.
23395524-540x390.jpg
A comprehensive portfolio of alkalis suitable for many analytical and R&D applications, with guaranteed specifications that conform to ACS and Reag. Ph. Eur. standards.
VWR_BDH_Chemicals_540x390.jpg
Choose from our alkalis for general laboratory use.
VWR_BDH_Chemicals_540x390.jpg
We offer a wide range of alkalis with limited specifications for general laboratory use.
VWR_BDH_Chemicals_540x390.jpg
We provide a complete range of salts suitable for a multitude of analytical and R&D applications. Our specifications are guaranteed to conform to ACS and Reag. Ph. Eur. standards.
ACS-and-Reag-3-540x390.jpg
Discover the advantages of our salts for general laboratory use.
23384486-540x390.jpg
We offer a wide range of salts with limited specifications is for general laboratory use.
22813638-540x390.jpg
See our comprehensive and wide range of "ready-to-use" tablets to meet the optimal sulfuric acid decomposition phase requirements.
12826034-540x390.jpg
For quality control in pharmaceutical applications, we offer more than 250 standards and solutions that conform to current European pharmacopeia standards. Each is delivered with a complete CoA to ensure complete traceability.
vwr_chemical_titration-540x390.jpg
To help you with your future titrations, find our complete range of pH, metal and redox indicators.
Lab auxiliaries, safety products and detergents
64571-540x390.jpg
Avantor® offers an extensive selection of products for ensuring safe lab operations and managing spills, including the versatile Algosol and basic spillage absorption granules.
VWR_BDH_Chemicals_540x390.jpg
Analytical-grade pumice stone granules are available from VWR® Chemicals.
40109762-540x390.jpg
We can provide industrial sands and other products for use in clarification processes.
Molecular-3-540x390.jpg
Multiple molecular sieves are available, with different materials to adsorb molecules with specific critical diameters.
43237260-540x390.jpg
These silicone grease lubricants are available for use with a variety of lab glassware.
VWR_BDH_Chemicals_540x390.jpg
A variety of gases are available for purification tasks.
VWR_BDH_Chemicals_540x390.jpg
These standard freezing products are used for a variety of lab tasks.
Silica-1-540x390.jpg
The VWR® Chemicals portfolio includes multiple drying agents to clean up spills.
IMG_9327-540x390.jpg
LABWASH® offers a range of manual or machine washing products for housekeeping tasks.
40109803-540x390.jpg
Our offerings include Chameleon® absorbent granules and non-toxic desiccants for adsorbing moisture from the atmosphere and gas streams.
43217776-540x390.jpg
Our silicone fluids are low in toxicity, inert and long-lasting due to their low volatility. They are recommended for use in baths.
121-0006-540x390.jpg
Our products include convenient spill kits and absorbent granules.