ContactUsLinkComponent
mabs-env-control-522549218-1920x760.jpg
mAbs Formulation and Final fill 
Progressing from the bulk drug substance to the final injectable drug product requires products like our J.T.Baker® line of excipients including surfactants, stabilizers, and bulking sugars. Avantor products meet quality and performance specifications that ensure product stability, solubility, and bioavailability.
ISO-certified environments critical to final fill
To meet final fill requirements, products must be prepared in an ISO Class 5 environment and meet USP71, USP85, and USP758 requirements to be sterile, pyrogen-free, and particulate-free.
They must also be validated for use in storage of frozen or lyophilized drug product and for compatibility with the protein and excipients in final formulation.
We offer a range of excipients that are used in drug product formulation – including adjuvants, stabilizers, preservatives and bulking agents – as well as final packaging vials, and stoppers and other critical materials for final fill step. Our production water used in drug product manufacturing and as a diluent, which is processed and meets USP Water-for-Injection (WFI) testing requirements.
Explore product choices for final fill for mAbs
production-biochemicals-801080266-540x390.jpg
With cGMP and cGDP chemical and excipients manufacturing, see how Avantor’s offerings can help speed time-to-market.
single-use-530817724-540x390.jpg
Discover Avantor's fluid handling solutions and access literature, technical articles and videos.
bioprocess-support-2-540x390.jpg
Learn how Avantor Services bioprocessing specialists can help optimize your upstream and downstream operations.
env-control-184113244-540x390.jpg
Avantor’s environmental control solutions include our complete product line and CE specialists with ISO and IEST protocols training.
Explore product choices for final fill for mAbs
monoclonal-antibodies-260x190.jpg
J.T. Baker® Amino Acid range
De-risk your supply chain with Avantor’s US country-of-origin and ChP compliant J.T.Baker® Amino Acid range.
Contact us
Related content
vaccination-injection-1283486156-540x390.jpg
Arvind Srivastava, Technical Fellow; Ger Brophy, EVP, Biopharma Production Solutions and Meera Agarkhed, former Sr. Manager, Technology and Innovation discuss how formulations containing citrate compare to other buffers in reducing SC injection-site pain in a peer reviewed paper.
Ready to maximize production efficiency?
Our Bioprocessing Solutions team is here to help.
Yes, let's get started!