ContactUsLinkComponent
Cleanroom Stationery
basan_Statio_StickyPads_200.jpg
Cleanroom notebooks and cleanroom sticky pads are made from cleanroom paper. Writing on both sides of the sheet is possible. We recommend our durable and resistant clipboards to facilitate the scribble of notes in your cleanrooms and controlled production areas.
Special cleanroom pens with a non-slip grip ease writing even with cleanroom gloves. The ink (different colours are available) of the cleanroom pens is permanent, even on cold and wet surfaces (e.g. plastic bags, disposable labware), and quick drying. It will not smear but can be scrubbed off.
Product selection