ContactUsLinkComponent
Disposable Cleanroom Garments
Disposable garments are designed and manufactured from lightweight, low-linting and durable material, providing an excellent combination of both comfort and protection.
basan provides the full range of disposables for cleanrooms and controlled environments, covering the full head-to-toe range, including overalls, coats, hoods, sleeve protectors and boot covers. To suit all areas, the range is available in sterile and non-sterile.
basan_Disp_Graphik_P22_EN.jpgbasan_Disp_Fabrics_P22_EN_530.jpg
Product selection
basan_Disp_Coverall_200.jpg
With hood attachment, with integrated socks or as sterilisable version.
basan_Disp_Coat_200.jpg
Made of different materials, lengths, with press studs or velcro fastener, with/without hip pocket.
basan_Disp_Apron_200.jpg
Tyvek, LPDE, Chemotherapy protective or chemical resistant aprons available.
basan_Disp_Hood_200.jpg
Available as nonwoven hoods, Tyvek hoods or peaked caps.
basan_Disp_Beardcover_200.jpg
Nonwoven beard covers with good barrier ability against particles. Breathable, universal size.