VWR® CHF5CB coveralls
Don´t compromise on comfort or safety
Are you looking for a good basic protection for applications that are less demanding in terms of protection? These VWR CHF5CB overalls are not hot to wear or as fragile as microporous film overalls. They offer more protection than SMS overalls and meet your budget requirements.
- Cat III - Type 5/6 chemical protection
- Softer feel: Touch the garment and feel the difference
- Different balance of protection and comfort
- Based on a new lower cost polytheylene non woven fabric
- Optimised design and packaging
| Clothing size | Pk | Cat. No. |
|---|---|---|
| S | 25 | 113-8379E |
| M | 25 | 113-8380E |
| L | 25 | 113-8381E |
| XL | 25 | 113-8382E |
| XXL | 25 | 113-8383E |
| 3XL | 25 | 113-8384E |