Orders LinkContactUsLinkComponent
OvensAndIncubators-955x690.jpg
Ovens and Incubators
VWR® ovens and incubators from Avantor® provide superior temperature accuracy and excellent recovery time for a wide range of demanding applications. Forced air mechanical convection units offer uniform heating, precise temperature control and fast processing using a blower that circulates heated air in a horizontal airflow pattern. Gravity convection units feature a vertical airflow pattern perfect for drying samples that require a low-turbulence environment.
Ovens & Incubators | Avantor
Digital advanced and adaptive microprocessor control systems provide superior temperature accuracy. All units feature a durable, powder-coated steel exterior with stainless steel interior; glass wool insulation; and high-temperature grade, foamed silicone rubber door gasket.
CarbonDioxideIncubators-400x495.jpg
Ideal for busy laboratories where contamination concerns and ease of cleaning are important factors.
FanAssistedAndNaturalConvectionOvens-400x495.jpg
Dry and heat your samples with precision. Browse our selection of forced air and gravity convection ovens.
LowTemperatureBODIncubators-400x495.jpg
Offer excellent temperature uniformity for a wide range of applications.
VacuumOvens-400x495.jpg
Suitable for drying, baking, conditioning, out-gassing solids and liquids, vacuum embedding, moisture testing and age testing.
ForcedAirAndGravityConvectionIncubators-400x495.jpg
Provide a stable environment for your samples with our selection of forced air and gravity incubators.
ConstantClimateIncubators-400x495.jpg
Test chambers for constant climatic conditions provide consistent temperature and humidity values for long-term testing including shelf-life verification and packaging testing.
CooledAndPlantGrowthIncubators-400x495.jpg
Great for seed cultivation, fruit fly culture, protein crystal growth, BOA testing and more.
DryingOvensWithNaturalConvection-400x495.jpg
For routine drying applications and thermal testing.