Orders LinkContactUsLinkComponent
Mettler-Toledo-Hero-Block.png
pH and Conductivity Measuring Solutions for Laboratory and Field Applications
METTLER TOLEDO is a leading global manufacturer of precision instruments and services for use in laboratories and manufacturing. Lab pH instruments, pH and conductivity measuring solutions for laboratory and field applications, and electrochemistry systems provide accurate measurements of pH, conductivity, ion concentration, ORP/redox, and dissolved oxygen. To complete your measuring solution, you can pair our meter with one of our high-quality sensors, calibration solutions, or analytical software.
Mettler Toledo | Avantor
Lab Electrochemistry Instruments
Electrochemistry systems provide accurate measurements of pH, conductivity, ion concentration, ORP/redox, and dissolved oxygen for laboratory and field applications. To complete your measuring solution, you can pair our meter with one of our high-quality sensors, calibration solutions, or analytical software.
NineFocus meters
NineFocus™ is a high-performance, multiparameter benchtop meter for compelling precision and unlimited possibilities in pH, conductivity, ion concentration, redox, and dissolved oxygen determinations.
SevenDirect.png
The SevenDirect™ are accurate and flexible meters for the determination of pH, ion, and conductivity measurements.
FiveEasy.png
Thanks to the FiveEasy™ intuitive operation and sleek design, you can maximize your focus on pH and conductivity, instead of dealing with complex systems.
Seven2Go.png
These robust IP 67 meters provide durability and one-handed operation, making them ideally suited for mobile applications in the laboratory, at-line or outdoors.
FiveGo.png
These robust IP 67 meters deliver reliable measurements at the push of a button – ideal even in harsh conditions.
Electrodes.png
Measure pH, conductivity, temperature, ORP, and more with our wide range of electrodes. Find the right electrode for your lab today.
BuffersStandards400-495.jpg
Calibrate and validate your electrochemistry measurements with this extensive range of solutions.
Densitometers
Measuring the density and specific gravity at a given temperature, density meters are especially useful when working with viscous liquids or difficult gases. They also offer large internal storage capacity, allowing you to save and compare a high volume of sample data with ease. Used in various research or product-developing industries to check batch consistencies, these digital devices guarantee quality control.
Densito.png
Designed to fit comfortably, the lightweight Densito™ allows for one-handed operations in the lab or on the go.
Would you like more information?