Orders LinkContactUsLinkComponent
i-Thermo_M5_comp_f.jpg
Moisture analysis
Moisture analysis is a critical component of quality control, production control, and product development in many industries, including pharmaceutical and food. Moisture levels can influence product shelf life and impact other applications in many ways. Moisture analyzers provide a rapid alternative to traditional oven drying methods.
Moisture analysis | Avantor
Avantor® offers a comprehensive selection of moisture analyzers from top brands such as Sartorius, OHAUS and our VWR® brand. Each analyzer is designed with specific features to meet distinct application requirements. Among those features are infrared or halogen heating for expedited results and ceramic or coiled quartz elements for precise measurements.
Verified versions (OCE) are available for use in legal metrology.
Product selection
A wide range of moisture analysis products for every application 
i-ThermoG_VWR-540x390.jpg
An economical range of rugged, compact instruments for routine measurement.
MB32_right_open_without_sample-540x390.jpg
OHAUS moisture analyzers focus on accuracy and ease of use in routine and professional tasks.
wl-10405-e_wb_RGB-540x390.jpg
Premium quality with a focus on precision, for routine and professional use.
611-1374-540x390.jpg
Weighing and drying dishes
Sturdy aluminum dishes that are compatible with most moisture balances and dryers.
Have questions about our moisture analysis products?