Orders LinkContactUsLinkComponent
903850_Kabeltrommel-KT-MPP-SK_feature.jpg
Intelligent Digital Sensors (IDS) for measuring under harsh environmental conditions, WTW, a Xylem Brand
Intelligent Digital Sensors (IDS) for measuring under harsh environmental conditions, WTW, a Xylem Brand | Avantor
MPP IDS – the digital multi-parameters (up to three main parameters) depth probes with built-in depth measurement to use in profile measuring:
  • Depth measurement of pH, conductivity, dissolved oxygen, turbidity - up to three main parameters simultaneously, including depth and temperature measurement
  • Barometric pressure compensated depth measurement for accurate results
  • Profile measurement without a cable mess - special cable reel with sliding contacts available
  • New Plug Head System
  • No twisting, no screwing: Only a click – and a pressure, tension resistant, data proof connection to the MPP IDS is made. With thin and tear resistant cables in different lengths.
Typical applications include limnological testing of waters with a depth of up to 100 m, but also measurements in bore holes, monitoring of landfills and more. The Multi 3630 is required to measure for all MPP IDS.
ids-harsh-env-conditions-image.jpg
Depth profile and water quality measurement with the multi-parameter probe MPP IDS:
  • Easy configuration
  • Measurements down to 100 meters
  • Safe data transfer with USB flash drive