Orders LinkContactUsLinkComponent
AtomicSpectroscopyImgnew.jpg
Atomic Spectroscopy
Atomic spectroscopy is an advanced spectroscopic technology that analysts use to determine the composition of sample materials or objects in commercial, government, research and academic labs. Avantor® carries a comprehensive selection of spectrometers and other equipment and materials you’ll need to successfully determine the atomic identity of samples in your laboratory.
Atomic Spectroscopy | Avantor
undefined
Questions about our atomic spectrometry products and services?
Contact us today to discuss your needs. Our team is ready to help.
Contact us
Featured Suppliers
1-99001-0001-540-390.jpg
Quality control of ICP Standards by an accredited calibration laboratory according to ISO 17025
SpecpureAnalyticalStandardsImgnew.jpg
Thermo Scientific offers a wide range of spectrochemical analytical standard solutions. The Specpure® analytical standards are manufactured for exceptional performance in accuracy, purity and quality. Browse the range today.
Closed-Bags-Together-540x390.jpg
VWRC-BDH ARISTAR ICP and ICP/MS Certified Reference Standards in enhanced packaging specially designed to prevent transpiration and ensure extended shelf life.
NL0229-540x390.jpg
These sample cups are designed to eliminate the need for trimming extraneous thin-film sample support material and offer top sample loading.
What is Atomic Spectroscopy?
By applying electromagnetic radiation to a compound, it puts atoms in the sample into an excited state. As they return to their original condition, they emit energy as light. By measuring the wavelength and intensity of the emitted light, analysts can identify the atoms in the sample.
Are you ready to move your next innovation forward? We can help.