Orders LinkContactUsLinkComponent
Gas Regulators
Gas cylinders provide the gas used to drive equipment for a number of laboratory applications. To regulate the delivery of the gas from the cylinder, the proper regulator selection is required. Whether your application requires high-purity, ultra-high purity, corrosive or blended gas, Avantor® offers all the single-stage and multi-stage regulators you need for every application.
SF_1152_blck1.jpg
VWR General Purpose Single-Stage Gas Regulator
  •   Applications where small changes in gas delivery pressure will not degrade performance
  •   Acetylene AA / FID / ICP (VWR # 55850-200)
  •   Air ICP (VWR # 55850-260)
Learn More
SF_1152_blck2.jpg
VWR High-Purity Single-Stage Gas Regulators, Brass
  • Applications where pressure rise is not critical
  • Calibration Gases, Process Analyzers, Gas Chromatography (GC), EPA Protocol mixes
Learn More
SF_1152_blck3.jpg
VWR High-Purity Single-Stage Gas Regulators, Stainless Steel
  • Applications where a rise in delivery pressure from a full to empty cylinder can be tolerated
  • Ultra- add High-Purity and Corrosive Gas for Process Analyzers, Gas Chromatography (GC), EPA protocol mixes, laser gas systems and emission monitoring systems
Learn More
SF_1152_blck4.jpg
VWR General Purpose Multistage Gas Regulator
  • Applications requiring constant delivery pressure with no fluctuation
Learn More
SF_1152_blck5.jpg
VWR Multistage Gas Regulator
  • Applications where constant working pressure is important over a wide range of inlet pressures and flow rates
  • Gas Chromatography (GC)
  • CO2 Incubators (VWR # 55850-414)
Learn More
SF_1152_blck6 .jpg
VWR High-Purity Two-Stage Gas Regular
  • Applications requiring constant delivery pressure regardless of change in cylinder pressure
  • GC and GC/MS
  • Helium/Nitrogen (VWR # 55850-624)
  • Hydrogen (VWR # 55850-632)
  • Zero Air (VWR # 55850-642)
Learn More
SF_1152_blck7 .jpg
VWR High Purity Two-Stage Gas Regulator - Stainless Steel
  • Applications needing constant outlet pressure regardless of change in cylinder pressure
  • Corrosive gas applications
Learn More
SF_1152_blck8 .jpg
Regulator Accessories
VWR Semiautomatic Switchover Manifolds
  • Designed to prevent depletion of gas during testing and eliminating downtime required to change empty cylinders.
  • CO2 Incubators (VWR # 82023-736)
Learn More
SF_1152_blck9 .jpg
  • Ideal for use with cylinders with maximum inlet pressure of 3000psig
  • For use with high-purity noncorrosive and corrosive gases
  • Ideal for CO2 incubators