Orders LinkContactUsLinkComponent
400113Featurecardimg.jpg
Quick Imaging and Analysis
Quick Imaging and Analysis | Avantor
Applications
  • Cell counting and cell viability
  • Assessing mito chondrial and cytotoxicity
  • Measuring marker expression
  • Measuring neurite outgrowth
Product Details
The SpectraMax MiniMax 300 Imaging Cytometer option is a field-upgradeable option for the SpectraMax i3x Multi-Mode Detection Platform  that offers cellular imaging capability.
The MiniMax 300 enables quick imaging and analysis of cells, and allows this to be combined with absorbance, luminescence, fluorescence and Western Blot assays for a complete, end to end solution.
Request a Demo