ContactUsLinkComponent
WesternBlottingHeroPage.jpg
Protein Electrophoresis and Western Blotting
Getting accurate and high-quality results depends on having the supplies and equipment you need to do your job efficiently and effectively. That’s why Avantor® carries your preferred products from brands you trust.
undefined
Learn more about our protein electrophoresis and western blotting solutions.
Contact us today to discuss your needs. Our team is ready to help.
Contact us
Featured Suppliers
Featured-Suppliers.jpg
What’s the most efficient way to do total protein normalization of NIIR blots? With the Sapphire™ Biomolecular Imager from Azure Biosystems.
Cytiva.jpg
Cytiva provides the tools that help advance and accelerate the development and manufacture of therapeutics.
Protein Extraction & Sample Prep
Sample-Prep.jpg
Quality data depends on high-quality sample preparation. Find everything you need to prepare high-integrity protein samples and successfully extract them.
Protein Electrophoresis
Transfer
Probing & Imaging
What is Western Blot?
Western-Blot.jpg
Western blot, or western blotting, is a process that detects specific proteins in a complex mixture of proteins extracted from cells or tissues.
Western blot originated in the late 1970s and is now a routine protein analysis technique used extensively by biochemists, molecular biologists and cell biologists. The method results in both qualitative and semi-qualitative data and is helpful in a wide range of applications, including:
  • Lyme disease ·
  • BSE (Bovine Spongiform Encephalopathy)
The three main steps in the workflow are separation by size, transfer to stable support and visualization using appropriate primary and secondary antibodies.
Step one: Separation by size
Scientists and technicians use protein gel electrophoresis to separate protein mixtures according to the molecular weight of each component protein.
Step two: Transfer to stable support
After the proteins have been separated, they are transferred to a stable support membrane using a blotting apparatus.
Step three: Visualization with primary and secondary antibodies
In step three, antibodies specific to the protein of interest bind and are detected after the film is developed or digital images are processed.
How to Choose Your Western Blot Equipment
Blot-Equipment.jpg
Reliable results may depend on using the proper western blotting protocol specific to your western blot equipment. Blotting kits simplify the purchasing process and include all necessary components to transfer protein, DNA, or RNA samples onto membranes for staining.
Avantor® can deliver individual components from various equipment, products and supplies to help create a solution that supports your workflow.
What Is Protein Electrophoresis?
Western-blot2.jpg
Protein electrophoresis is a laboratory technique in which charged protein molecules are separated according to physical factors by an electrical current that forces them to move through a gel. Protein gel electrophoresis is used in many proteomic analyses, including western blot and helps identify various diseases. Protein electrophoresis is used to diagnose a type of cancer called myeloma, or multiple myeloma. It can also be used to diagnose other conditions, including:
  • Waldenström macroglobulinemia
  • Primary amyloidosis
  • Monoclonal gammopathy of undetermined significance (MGUS)
  • Diabetes
  • Anemia
  • Thyroid problems
  • Liver diseases
  • Certain autoimmune diseases
  • Nutrient absorption inability or poor nutrition
Selecting the Right Protein Gel for Your Research Application
Protein1-Electrophoresis.jpg
Select the gel that’s best for your protein electrophoresis experiment by matching gel features to your needs. Consider your requirements for the following options:
  • Sample type
  • Gel format
  • Compatible gel tank
  • Gel chemistr
  • Sample volume
  • Separation type
  • Well format
  • Separation range
  • Number of wells
Featured Products
Are you ready to move your next innovation forward?
We can help. Contact us to speak with a member of our team.
Contact us