Orders LinkContactUsLinkComponent
PCR Sample Protection | Avantor
PCR Sample Protection
pcr-sample-540.jpg
The VWR® PCR Workstation Pro HEPA offers twin decontamination action by UV inactivation of airborne and surface-bound contaminants and, therefore, represents an ideal environment for PCR sample preparation and other sensitive protocols.
Additional Resources
More information