Orders LinkContactUsLinkComponent
NGSHero1920.jpg
Next Generation Sequencing (NGS)
Next Gen Sequencing is an umbrella term describing a collection of high-throughput methodologies and platforms used for the determination of the sequence of base pairs in DNA or RNA.
Next Generation Sequencing (NGS) | Avantor
undefined
Questions about our NGS product offering?
Contact us today to discuss your needs. Our team is ready to help.
Contact us
Featured Suppliers
NEB950.jpg
New England Biolabs (NEB): Shaping the Landscape of Bioscience Research
NEB offers a broad portfolio of reagents designed to streamline RNA analysis and genome-editing workflows.
OxfordNanoporeTechnologies950.jpg
Oxford Nanopore offers many unique products tailored to improve scientific researchers’ NGS workflows.
Nucleic Acid Extraction & Purification
NucleicAcidExtraction540.jpg
The accuracy and reliability of research and diagnostic results depend on the initial materials used during sequencing. You can help obtain the maximum nucleic acid yield and purity by using extraction and purification products from Avantor®
Manual or automated benchtop homogenizers have user-friendly control systems for straightforward processing.
See our full offering of nucleic acid purification kits and Reagents.
PCR workstation enclosures provide the contamination-free environment required for the most sensitive DNA segment detection.
Get the automated nucleic acid purification systems you need for your lab.
Nucleic acid storage systems maintain DNA and RNA purity and quality for future processing.
Nucleic Acid QC & Storage
NucleicAcidQC540.jpg
Quality control is critical to ensure data is not affected by any contaminants. Browse the equipment we offer to help you quantify samples and maintain nucleic acid stability during storage.
View our offering of UV/Visible spectrophotometers.
Buffers and systems for nucleic acid stability during sample storage/transport.
Fluorescence spectrophotometers for a variety of research applications.
Library Construction
LibraryConstruction540.jpg
Find all the tools you need to efficiently prepare your nucleic acid target, RNA, or DNA for the specific sequencing system your research requires.
Kits for DNA fragmentation and library preparation for NGS.
Kits for DNA end repair and ligation of DNA adaptors for NGS.
Reagents and kits for reaction cleanup and size selection in NGS.
Find a selection of Hi Fidelity DNA Polymerases
See all the PCR Cyclers we offer.
PCR workstation enclosures provide the contamination-free environment required for the most sensitive DNA segment detection.
Library Quantification & QC
LibraryQuantification540.jpg
Avantor delivers accurate quantification and reliable quality control you can depend on to ensure successful sequencing.
Fluorescent dsDNA binding dyes to simplify DNA quantitation.
See all the qPCR Cyclers we offer.
qPCR kits for the quantification of NGS libraries
What is Next-Generation Sequencing (NGS)?
whatisNGS540.jpg
Next-generation sequencing (NGS) describes several techniques used to determine the sequence of DNA or RNA. Scientists can use that sequencing data to study genetic variations or mutations associated with diseases or biological conditions.
Because NGS allows scientists and researchers to sequence thousands or even millions of DNA molecules simultaneously, it has significantly reduced the cost and the amount of time needed to sequence genes and genomes. That has led to dramatic advances in clinical diagnostics, genetic diseases and personalized medicine.
Sanger Sequencing vs. Next-Generation Sequencing
SangerSequencing540.jpg
Sanger sequencing and traditional next-generation sequencing (NGS) are similar in concept, . Both techniques involve using DNA polymerase to add fluorescent nucleotides one at a time onto a growing DNA or RNA template strand. The fluorescent tags allow the identification of each successively incorporated nucleotide.
Where these two sequencing methods differ is in each method’s sequencing volume. The Sanger method sequences DNA or RNA one strand at a time, while next-generation sequencing – initially called “massively-parallel sequencing” – can simultaneously sequence hundreds to thousands of genes at one time.
More recently, new NGS methodologies have emerged. Often referred to as third-generation sequencing, these techniques enable the analysis of much longer DNA and RNA fragments than possible using either Sanger or traditional NGS – delivering significant advantages for a range of applications, including genome assembly and the analysis of large genomic aberrations (e.g. structural variants and repeats). One such technique, nanopore sequencing, enables the analysis of any length of DNA/RNA fragment – from short (20 bp) to ultra long (>4 Mbp). Furthermore, as amplification is not required, nanopore sequencing eliminates amplification bias and allows direct analysis of base modifications (e.g. methylation) alongside the nucleotide sequence While Sanger sequencing is still used frequently in clinical research labs, the use of NGS in these labs is expanding rapidly through the facility to cost-efficiently analyze a greater number of targets and samples at higher sensitivity .
Advantages of NGS
AdvNGS540.jpg
Next-generation sequencing provides multiple benefits when compared to Sanger sequencing, including:
  • Can sequence an entire genome or high numbers of targets at the same time
  • Faster turnaround time 
  • More cost-effective
  • Requires lower sample input
  • Improved accuracy and reliability
  • Higher sensitivity allows detection of variants at lower frequencies
  • Assay a greater range of variation in a single run using nanopore sequencing (i.e. SNVs, SVs, repeats, phasing, methylation)
Typical NGS Workflow
TypicalNGS.jpg
Traditional NGS platformNanopore sequencing
1. Library preparation includes fragmenting DNA or RNA from the rest of the sample, preparing DNA or RNA for sequencing by adding adapters and checking all fragments for size and quality before adding them to the sequencing library1. Attach sequencing adapters to DNA or RNA sample – no fragmentation required, enabling sequencing of any length of molecule, including ultra-long fragments (up to 4 Mb) for enhanced genome assembly and analysis of structural variants, repeats, phasing and transcript isoforms.
2. Clonal amplification generates multiple copies of target areas of interest within a DNA or RNA sample to ensure they provide a robust and detectable signal during the sequencing process.2. Load sample to Flow Cell containing hundreds of nanopores (nano-scale protein holes embedded in an electrically resistant membrane). No DNA/RNA amplification is required, eliminating amplification bias and enabling simultaneous identification of base modifications (e.g. methylation)
3. Sequencing is the process that tells scientists what genetic information each DNA or RNA molecule contains.3. The DNA or RNA molecules pass through the nanopores, disrupting the ionic current and generating distinctive signal that is decoded to provide the specific DNA or RNA sequence. All sequence data is provided in real-time, enabling immediate sample insights and actionable results.
4. Data analysis is the process of applying a wide range of analytical methods to DNA or RNA sequencing information to determine its structure, function, features, or other characteristics.4. Data can be analysed by a wide range of standard tools, including Oxford Nanopore’s EPI2ME platform, which offers a range of optimised best practice analysis pipelines
Are you ready to move your next innovation forward? We can help.