Orders LinkContactUsLinkComponent
petri-dish-1190512184-1920x760.jpg
New microbiology products
Avantor® continually invests in and expands its portfolio of products for the entire microbiology workflow, from sampling and sample preparation to the identification of microorganisms. That’s because the methods used to control microorganisms, their growth and their transmission never stop evolving.
New microbiology products | Avantor
undefined
Contact us for assistance.
Talk to us about your microbiology laboratory challenges. We can recommend new products and solutions to meet your needs.
Contact us
Choose from the latest innovative microbiology tools.
39308871-540x390.jpg
VWR® MPP15: simplifying culture media preparation
Simplify and accelerate culture media productions with the VWR® Medium Preparator Premium 15 (MPP15). Media can be effectively mixed, heated, sterilized and quickly cooled to the desired temperature. For filling, media are kept stable at the selected temperature. The MPP15 can also be used for autoclaving and sterilizing small objects and liquids in separate containers or as a water bath.
Additional information
35017297-540x390.jpg
VWR® automatic colony counters
Avantor® helps save time in your lab with the VWR® automatic colony counter. Automating this process delivers consistency, accuracy, speed and efficiency. Our new range of automatic colony counters includes everything from low-cost units for the rapid counting of pour plates, spread plates and spiral plates to high-end units for extended applications and IQ/OQ.
Additional information
T-Lab_Eco_540x390.jpg
T-Lab Eco: the economical laboratory autoclave
The T-Lab Eco offers the simplicity and economy many educational institutions and research facilities seek. This vertical floor-standing autoclave fully supports general lab sterilization requirements and helps increase lab productivity by combining the highest universal capacity with the optimization of resources such as water, power and processing time.
Additional information
29011699-540x390.jpg
CytoFLEX flow cytometer
Improve your enumeration assays with the CytoFLEX flow cytometer. The platform's advanced optics and fluidics let you resolve small particles down to 200 nm and count bacteria as small as 0.5 µm without the need for counting beads. It also provides information on viability and vitality of cells to save manufacturers time and money.
Additional information
36296484-540x390.jpg
Rapidly identify and fully characterize microorganisms from culture or complex metagenomic samples using affordable, portable MinION™ sequencing devices. They offer convenient, real-time analysis for rapid access to actionable results.
Additional information
Talk to your specialist
28763171-540x390.jpg
VWR® lockable contact plates
VWR® lockable contact plates feature a triple-bagged and lockable screw-on lid that is well-suited for use in pharmaceutical lab applications. Packed in bags of five plates, they are ideal for environmental control of air, surfaces, gloves and overalls inside restricted areas like isolators and cleanrooms.
Additional information
Discover the latest innovative tools for microbiology workflows from Avantor®.