Orders LinkContactUsLinkComponent
High-Throughput Photometry | Avantor
High-Throughput Photometry
Featured Solutions
tunable-.jpg
Compact and tunable absorbance microplate readers, the SpectraMax® ABS and SpectraMax® ABS Plus Microplate Readers bridge the gap between the affordability of filter-based readers and the flexibility of monochromator-based systems.
SpectraMax® ABS
Additional Resources
SpectraMax ABS® Plus
multiskansky.jpg
Multiskan Sky microplate and cuvette spectrophotometer is designed to be exceptionally convenient and easy to use for virtually any photometric research application, especially DNA, RNA, and protein analysis.