Orders LinkContactUsLinkComponent
Gene-expression-hero.png
Gene Expression Analysis & Genotyping Workflow
Genetic variations in genomes and gene expression patterns analysis are essential to drug discovery and development of therapeutics. Biomarker discovery and disease molecular signatures are essential for improved diagnostics.
End to end workflow solutions - Through partnership with leading suppliers, Avantor provides unmatched convenience supporting the evolving needs of the scientific community to find innovative solutions to the critical questions in genomics.
Gene Expression Analysis & Genotyping Workflow | Avantor
undefined
Questions about our solutions for gene expression analysis and genotyping workflow?
Be contacted by our Specialists to discuss about your needs.
Contact a Specialist
Featured content
OxfordNanoporeTechnologies-540x360.png
Solutions to enable the analysis of DNA and RNA. Discover how Oxford Nanopore can help to improve your NGS workflow.
gene-CA4.jpg
In this magazine you will find technical articles about innovation, expertise and solutions that will help you to succeed in your genomics challenges.
Learn more
Lifescienceresearchworkflow-gene-540x360.png
End to end workflow solutions. View our featured workflows and learn more about the breadth of available solutions to support your life science research needs.
Selected products & resources
Mag-Bind-400x495.png
Scalable and automatable cfDNA isolation kit, designed for rapid and reliable isolation of circulating DNA from 500 to 10000 µl plasma or serum samples. Can be processed manually or with automated platforms.
cDNA-synthesis-400x495.png
Faster, better, tougher next generation cDNA SuperMix for cDNA synthesis. Enhanced thermostability and inhibitor resistance. Superior speed – 10 min. reaction time (13 min. total protocol time)
Benchtopcellsorter-400x495.png
Bench top cell sorter, CytoFLEX SRT
Approachable instrument for sorting multiple populations to generate high quality material for downstream assays.
FragmentAnalyzerSystem-400x495.png
Fragment Analyzer System, 5300
Parallel capillary electrophoresis system used for nucleic acid QC in medium to high-throughput labs.
Sample Isolation & Purification
SampleIsolation-590x390.png
Upstream solutions to isolate and purify nucleic acids from samples prior to gene expression analysis and genotyping.
Manual or automated benchtop homogenisers have user-friendly control systems for straightforward processing.
See our full offering of nucleic acid purification kits and Reagents.
Get the automated nucleic acid purification systems you need for your lab.
View our offering of UV/Visible spectrophotometers.
Colony picker systems for clone screening and selection.
Robust, compact, high-value microplate and multimode microplate readers for all your biological assays.
Affordable flow cytometers and cell sorter to analyze cell populations.
Assay Preparation
Assa-Preparation.png
Solutions needed to set up/prepare the assays to analyse the samples.
Automated electrophoresis systems for nucleic acid sample QC and analysis.
See our selection of automated pipetting systems and automated liquid handling workstations.
Tips compatible with automated liquid handling platforms.
We offer a variety of End-point PCR reagents and kits.
Get all the reagents you need for qPCR.
Reagents and kits for Reverse Transcription PCR and cDNA synthesis.
We offer an extensive variety of PCR Plates.
PCR workstation enclosures provide the contamination-free environment required for the most sensitive DNA detection assays.
Kits and reagents for the preparation of DNA/RNA libraries for NGS.
Sample Processing & Analysis
SampleProcessing-column.png
Equipment used to analyze the samples.
See our thermal cyclers for PCR and qPCR applications.
See our range of nanopore DNA/RNA sequencing devices.
Robust, compact, high-value microplate readers for all your biological assays.
Systems delivering highly reproducible, quantitative data.
What is gene expression analysis?
Gene-expression-analysis-540x360.png
Gene expression analysis is the study of how genes are activated or deactivated to produce functional gene products such as proteins or RNA molecules within a cell. It involves examining the patterns and levels of gene activity to understand how genetic information is utilized in different biological processes. Techniques like microarrays and RNA sequencing are commonly used to quantify and analyze the abundance of messenger RNA (mRNA) or other gene products, providing insights into the functional roles of genes, the regulation of cellular processes, and the identification of potential biomarkers or therapeutic targets in various biological systems.
What is genotyping?
Genotyping is the process of identifying genetic variations, such as single nucleotide polymorphisms (SNPs), insertions, deletions, or other mutations, in an individual's DNA. This analysis helps researchers and clinicians understand genetic differences among individuals or populations.
What are the techniques used for gene expression and genotyping?
Techniques used for gene expression analysis:
  • Microarrays: Microarrays allow the simultaneous analysis of the expression levels of thousands of genes. They involve placing DNA fragments representing different genes on a solid surface and hybridizing them with labeled RNA samples to measure gene expression levels.
  • RNA Sequencing (RNA-Seq): This technique involves sequencing the entire transcriptome, providing a comprehensive view of gene expression. It allows the identification of novel transcripts, alternative splicing, and precise quantification of mRNA levels.
  • Reverse Transcription Polymerase Chain Reaction (RT-PCR): RT-PCR amplifies and quantifies specific RNA molecules, providing a targeted approach to measuring gene expression. It is widely used for its sensitivity and specificity.
  • Quantitative PCR (qPCR): qPCR is a variation of PCR that enables the real-time monitoring of DNA amplification. It is used for accurate quantification of gene expression levels and is particularly valuable in validating results from high-throughput methods.
Techniques used for Genotyping:
  • Polymerase Chain Reaction (PCR): PCR amplifies specific DNA regions, facilitating the detection of genetic variations. It is commonly used for targeted genotyping of known polymorphisms.
  • Single Nucleotide Polymorphism (SNP) Genotyping Arrays: These microarrays are designed to analyze large numbers of SNPs simultaneously, providing a high-throughput method for genotyping.
  • Next-Generation Sequencing (NGS): NGS technologies can sequence entire genomes or targeted regions, offering a comprehensive approach to genotyping and allowing the identification of novel genetic variants.
What are the main applications for gene expression analysis and genotyping?
Gene expression analysis has diverse applications across biological and medical fields:
  • Disease Research and Diagnosis: Studying gene expression patterns helps identify genes associated with diseases, aiding in understanding their molecular mechanisms. This knowledge contributes to the development of diagnostic markers and potential therapeutic targets.
  • Cancer Research: Gene expression analysis in cancer cells can reveal aberrant patterns that distinguish cancerous tissues from normal ones. This information guides cancer diagnosis, prognosis, and the development of targeted therapies.
  • Drug Discovery and Development: Analyzing gene expression in response to drugs helps researchers identify how drugs influence cellular processes. This information is crucial in drug discovery, allowing the development of more effective and targeted therapeutic agents.
  • Personalized Medicine: Gene expression profiling enables the customization of medical treatments based on an individual's genetic makeup. This approach enhances treatment efficacy and minimizes adverse effects by tailoring interventions to a patient's specific genetic characteristics.
  • Biotechnology and Agriculture: Gene expression analysis is employed in the development of genetically modified organisms (GMOs) with desired traits, such as improved crop yield or resistance to pests. It also contributes to understanding the regulatory mechanisms in plants and animals.
  • Neuroscience: Studying gene expression in the brain helps unravel the molecular basis of neurological disorders and provides insights into brain development, function, and plasticity.
  • Environmental Monitoring: Gene expression analysis in organisms exposed to environmental factors helps assess the impact of pollutants, toxins, or other stressors on living systems, aiding in environmental monitoring and conservation efforts.
  • Stem Cell Research: Gene expression profiling is crucial in understanding the differentiation and developmental potential of stem cells. It guides the use of stem cells in regenerative medicine and tissue engineering.
Applications of Genotyping:
  • Association Studies: Genotyping is used in genome-wide association studies (GWAS) to identify genetic variants associated with diseases, traits, or drug responses.
  • Forensic Analysis: Genotyping is applied in forensic science for individual identification, paternity testing, and criminal investigations.
  • Population Genetics: Understanding genetic diversity within populations and identifying specific genetic markers help study human evolution, migration patterns, and genetic predispositions.
Are you ready to move your next innovation forward? We can help.
Contact us to speak with a member of our team.
Contact a specialist