Orders LinkContactUsLinkComponent
AT-BELL29-00-and-10-and-20-PRIMARY-955x590.jpg
Refractometry and polarimetry
Refractometry is a basic method for quantitative analysis, assurance and control in the food and beverage industry. Grape growers and the sugar industry frequently use refractometers for Brix testing sucrose levels in fruit. For more precise results, polarimeters are used to measure the optical rotation of sucrose solutions. Avantor® offers a range of tools and materials to meet your specific analytical and quality assurance requirements.
Refractometry and polarimetry | Avantor
A large selection of refractometry and polarimetry equipment for food, beverage and other industrial applications.
Product selection
Handheld_refractometers_400x495.jpg
Handheld refractometers
The portable, easiest and most cost-effective way to get a RI or Brix result.
Abbe_refractometers_400x495.jpg
For traditional refractometry measurements.
BELL19-00-10-20-Traditional-400x495.jpg
Digital refractometers
Modern and precise refractometers for reliable, accurate documentation with printouts and data storage.
Polarimeters_400x495.jpg
Polarimeters
Avantor® offers a range of standard and digital polarimeters to fit your specific needs.
Certipur_refractometry_ref_material_400x495.jpg
Reference materials
CertiPUR® reference material for refractometry to help ensure accurate results.
Have questions about our refractometry and polarimetry equipment?