Orders LinkContactUsLinkComponent
water-treatment-153095885-1920x760.jpg
Masterflex® Water System Treatment Pump & Tubing Solutions
Maintaining water quality is a perpetual challenge. Only around 1% of the world’s water supply is fresh, and only a fraction of that is potable.
Masterflex® Water System Treatment Pump & Tubing Solutions | Avantor
Chemical feed pumps make all the difference when it comes to water treatment. The chemicals involved can include virtually any chemical precursor or end-product. And Masterflex peristaltic, piston, and gear pump technologies are the ideal pumps for water treatment plants.
Our pumps:
  • Regulate feed rates
  • Release chemicals in real-time into reaction vessels, required in reactant feed applications
  • Accurately dispenses end-product into molds and containers
Come and see how Masterflex pump solutions can change the water treatment game.
Masterflex peristaltic pump uses
Masterflex peristaltic pumps are designed for:
  • Decontaminating ground water
  • Chemical drum sampling
  • Enzyme isolation
  • Chemical synthesis
  • Flow injection analysis
  • Greenhouse watering
  • Fertilizer applications
  • Pesticide delivery systems
  • Sewage/sludge analysis
  • Tree spraying
  • Water salinity analysis
  • Wastewater sampling
Masterflex water treatment applications advantages
What makes Masterflex a top choice for water treatment applications?
  • Broad range of tubing materials: Our tubing is compatible with a wide variety of industrial chemicals.
  • Noncontaminating: Fluid only contacts inside of tubing, eliminating contamination risk.
  • Easy cleanup: Cleaning involves simply changing tubing. IP66 washdown models are available .
  • Self-priming: Suction elevations up to 8.8 m (29 ft).
  • Handles viscous materials: Oils and other viscous materials are no problem. Transfer and dispense oils or fluids in suspension.
  • Runs dry: Pumps can be left unattended and won’t suffer damage when pumping dry.
  • Reversible: With a single pump you can purge and collect a sample.
  • Modular design: Mix and match interchangeable pump heads, tubing, and drives.
  • Custom engineered service: Fits right into your lab design without workflow disturbance.
Featured Masterflex water system pumps
mflx-77915-10-150.jpg
Ideal for chemical feed/metering and applications requiring high-purity, high-pressure or both.
mflx-07571-02-150.jpg
Portable Sampling Pump
Designed for the ultimate in portability and versatility. Run on rechargeable battery or plug in to AC or 12 VDC.
mflx-38705195-150.jpg
High-Volume Dispensing Pump
Durable, washdown, variable-speed digital drive can handle large batches quickly and efficiently with flow rates up to 42 LPM.
masterflex-masterflex-l-s-precision-pump-tubing-pl-150.jpg
Masterflex and Ismatec precision extruded and pump tested formulations engineered for the highest reliability, compliance, and traceability.
Discovering more water treatment pumps and solutions
Turn to Masterflex® for all your water treatment pump solutions.
Contact us today and discover what we can do for you at 1-800-MASTERFLEX or [email protected]