Orders LinkContactUsLinkComponent
mflx-single-use-components-1920x760.jpg
Masterflex® single-use components
Choose reliable components from Masterflex® adhering to strict sterilization protocols 
Masterflex single-use components | Avantor
Leverage the advantages of Masterflex single-use bioprocess assemblies
With Masterflex single-use, you will no longer have to validate the cleaning and sterilizing of your bioprocess assemblies. The purchase of a pre-assembled, sterilized and validated system shifts the burden of validation to the supplier. That is why it is important to be sure your supplier uses only the most reliable brand-name products and adheres to strict, well-established protocols in the sterilization process.
Masterflex single-use assemblies are manufactured in a Class 10000 cleanroom. The assembly facility is ISO 13485 certified and are cGMP compliant. We offer gamma irradiation sterilization with a 10–6 SAL (Sterility Assurance Level), a one millionth probability of microbial survival, and full lot traceability that identifies the production site, manufacturing date and origin of the components. We also provide and maintain all required documentation, including complete validation documents and certificates of conformance and sterility.
We build custom assemblies with reliable products:
  • Fittings
  • Tubing
  • Sensors
  • Common accessories
Fittings
masterflex-fitting-sanitary-fitting-with-fused-gas.jpg
Sanitary Fitting with Fused Gasket
Add another layer of safety to ensure sterility by preventing contamination with zero user contact with the gasket. No adhesives or clips needed to hold the gasket in place. 
masterflex-quick-disconnect-fittings.jpg
Quick-disconnect fittings allow you to make and break tubing connections without separating tubing from fitting. Available connections – barbed, pipe, compression and push-to-connect. 
masterflex-luer-fittings.jpg
Luer fittings are designed to work with microtubing and luer-type syringes. Standard-sized connecting ends make luer fittings, stopcocks and manifolds interchangeable.
masterflex-barbed-fittings.jpg
Barbed fittings slide into flexible tubing. Tubing elasticity holds the tubing onto the fitting for low-pressure installations.
Tubing
masterflex-puri-flex-200.jpg
  • Premium translucent tubing offering lower spallation and longer pump life than silicone
  • Heat-sealable and weldable
  • Ideal for sensitive cell work
masterflex-platinum-cured-silicone-200.jpg
  • Smooth surface; lower protein binding levels
  • Fewer potential leachables
  • Ideal for pharma and biotech use
masterflex-pharmapure-200.jpg
  • Popular opaque tubing option for bioprocess applications
  • Non-toxic, non-hemolytic, heat sealable, and weldable
  • Offers a long pump life 
masterflex-pharmapure-200.jpg
  • Biocompatibility similar to PharMed BPT®
  • Long life under continuous pressure up to 40 psi (2.7 bar)
  • Very low spallation and low extractables 
masterflex-chem-durance-bio-200.jpg
  • When you need chemical resistance and biocompatibility with extra low spallation, there is no better choice
  • Excellent life and durability under pressure
mflx-custom-cut-to-length-tubing.jpg
  • Choose from a wide selection of formulations
  • Save valuable time and money 
Sensors
masterflex-flow-sensors-200.jpg
  • Build a complete single-use flow sensor system customizable to your application
  • Ideal for single-use applications and suitable for long-term measurement
  • Non-invasive design eliminates contact with working fluid
mflx-pressure-sensor-200.jpg
  • Ideal for applications such as tangential flow filtration, cross flow filtration, depth filtration, bioreactor monitoring and chromatography
  • Sensors priced for single-use systems but may be reused
mflx-temperature-sensor-200.jpg
  • Eliminates cross-contamination and extensive cleaning protocols
  • Can be sterilized by gamma irradiation
  • Better than ±0.2°C accuracy
Common accessories
mflx-ratchet-clamp-200.jpg
Durable plastic clamp is autoclavable and will not rust, corrode or distort. Twelve-position ratchet control adjusts from full flow to complete closure. Accepts flexible tubing to ½” OD.
mflx-sanisure-clamp-200.jpg
SaniSure® Clamps
An innovative clamp that offers 360° compression for your sanitary connection. Meets USP Class VI requirements.
mflx-heavy-duty-tubing-closure-200.jpg
Durable clamp meets the working pressure of large tubing. High-strength polysulfone closure provides excellent durability that can be reused, and resists most chemicals.
mflx-barblock-200.jpg
Barblock
Achieve uniform 360° compression for a leak-free connection to barbed fittings. Provides seven times the pull resistance of cable ties without crimps or distortion.
It’s time to make your process more efficient
  1. What you need
    Choose from thousands of components. We custom configure, validate and package according to your specifications.
  2. When you need it
    Stop cleaning and revalidating your system. We manage your inventory and send assemblies according to your schedule.
  3. Call for a quote today
    Masterflex single-use experts provide a quote with full schematic, followed by an actual sample for your approval.
Find the single-use components you need from Masterflex®.
Contact our experts today to get started.
Contact us