ContactUsLinkComponent
mflx-IP-pump-series-1920x760.jpg
Masterflex® I/P® Series Peristaltic Pumps
Process pumping solutions
Powerful pumps built to last in challenging process and production environments.
For flow rates 0.0006 to 26 L/min
Masterflex I/P Series peristaltic pumps have powerful motors and better protection from production and industrial environments, making them ideal for process applications. Higher flow rates empower efficient fluid handling, with engineering designed for rigorous use.
I/P series pumps are used for:
  • Process and manufacturing
  • Media prep and hydration
  • Filtration and harvest
  • Medium-flow fluid transfer
  • Industrial production
  • Chemical sampling
Pumps are available with flexible options for monitoring and controlling your fluid handling, including Ethernet/IP, PROFIBUS, analog control, serial control, and MasterflexLive, which allows you to get real-time updates on your system. Peristaltic pumps are a low maintenance and cost-effective way to automate or expedite liquid handling in your processes. It’s easy to change tubing, with many options for tubing formulations to ensure compatibility with your process fluid.
Masterflex I/P Series Pump Features and Benefits
When you need nominal flow rates up to 26 LPM and pressures up to approximately 2.7 bar, our I/P series pumps are the ideal choice. Our pumps are:
  • Dependable and low maintenance, causing less disruption to your process
  • Designed for scalability- R&D to process
  • Minimize risk with a closed system, fluid only touches tubing
  • Handles transferring shear-sensitive and viscous fluids
  • Perfect for filtration, media feeds, bag filling, chemical sampling applications
  • Compact
All Masterflex I/P pump heads are designed to work best with I/P Precision or I/P High Performance precision pump tubing, ensuring lot-to-lot traceability and confidence in pump performance.   
The Masterflex MasterSense I/P pump features an intuitive touchscreen interface, which includes: 
  • Powerful drives that accept all I/P pump head and tubing options
  • Save commonly used dispense programs
  • Batch control
  • Volume dispense accuracy
  • Network connection via standard EtherNet/IP, Profibus, analog, or MasterflexLive
Masterflex pumps are built to last from high quality materials. Their precision engineering allows rigorous control over fluid volume and flow rates. And our digital MasterSense pumps include the latest in easy-to-use touchscreen technology, so you can get started right away.
Masterflex I/P pumps offer:
  • Nominal flow rates up to 26 LPM and pressures up to approximately 2.7 bar
  • Powerful drives that accept all I/P pump head and tubing options
  • Dependable and low maintenance, causing less disruption to your process
  • Designed for scalability - R&D to process
  • Minimize contamination risk with a closed system, fluid only touches tubing
  • Efficiently handles shear-sensitive and viscous fluids
  • Perfect for filtration, media feeds, bag filling, chemical sampling applications
  • All Masterflex I/P pump heads are designed to work best with I/P Precision or I/P High Performance precision pump tubing, ensuring lot-to-lot traceability and confidence in pump performance
    Featured I/P Pump Series Products
    mastersenes_drives_200x200.jpg
    Versatile pumps for process applications – with an intuitive touch-screen interface and advanced connectivity.
    RapidLoadPumpHead_200x200.jpg
    High-accuracy pumps with easy-to-change pump heads for fast and efficient tubing changes.
    BrushlessProcessDrives_200x200.jpg
    Powerful, low-maintenance drives that are cost-effective without sacrificing versatility and accuracy.
    Masterflex digital interface and MasterflexLive
    MasterflexLive® is a secure, cloud-enabled platform for monitoring Masterflex pumps; get real-time notifications of pump parameters including speed, flow rate, dispense volume, and more. Monitor critical processes running 24/7 without the requirement of onsite personnel - push notifications alert you to operating conditions and error messages.
    Masterflex I/P MasterSense digital pumps have an intuitive user interface with many programable options for your fluid handling workflows. Keep your fluid path solution up-to-date with over-the-air updates, so you always have the latest features and functionality.
    The Masterflex I/P MasterSense pumps feature an intuitive touchscreen interface, with features including:
    • 7'' (17.8 cm) capacitive TFT/RGB touch screen for crisp resolution and a wide viewing angle
    • Continuous run, volume dispense, and time dispense modes -- batch control and interval/off time in Volume and Time modes
    • Ability to save and retrieve commonly used dispense programs for rapid set up
    • Over-the-air firmware updates let you take full advantage of improvements in drive features and functionality
    • Options for connectivity and monitoring include MasterflexLive, Ethernet/IP, PROFIBUS, serial or analog signal
    • Three-level user access – gives you the option of controlling who can operate the pump and at what level
    • Real-time pressure sensing and alarms increase process reliability and frees up operator time
    Custom services and tools
    service-custom-eng-oem-100x100.jpg
    Partner with our OEM team and make your custom fluid handling idea a reality.
    custom-cut-tubing-service-100x100.jpg
    For best performance of your Masterflex pumps, use Masterflex tubing – choose type, length, and quantity.
    mflive-lp-100x100.jpg
    Remotely monitor your pump parameters in real time, from anywhere.
    bioconnect-service-100x100.jpg
    Set up disposable, sterile bioprocess assemblies with Masterflex quality systems.
    Masterflex I/P Series Pump Components
    Need more information or help selecting the perfect product for your application?
    Our experts are ready to help. Contact us at 1-800-MASTERFLEX or [email protected]
    Contact us