Orders LinkContactUsLinkComponent
Masterflex_I-P_MasterSense_Process_Drives_1920x760.jpg
Masterflex® I/P® MasterSense™ Process Drives
Reimage bioprocessing
NEW integrated sensor technology, intuitive touchscreen and MasterflexLive® remote monitoring for the biopharma industry of tomorrow
MasterSense™ monitors your media so you don't have to
With the extra time, a process engineer could:
MasterSense_monitors_your_media_so_you_dont_have_t.png
Advanced Connectivity and Control Options
Monitor from anywhere
connectivity-img-diagram-IP.jpg
MasterflexLive
  • MasterflexLive is our secure, cloud-enabled platform for real-time remote monitoring of all Masterflex pump parameters
  • Monitor critical processes running 24/7 without the need for onsite personnel
  • Push notifications alert you to changes in operating conditions and error messages
Learn more
Flexible Connectivity
  • Your choice of connectivity options-MasterflexLive®, EtherNet/IP, or Profibus and RS232 options
  • Three-tier access to pump functionality-use this option to protect your process by controlling who can operate the pump and at what level
  • Over-the-air firmware updates-automatically allows you to take full advantage of future enhancements
  • Internet connection via Wi-Fi or standard Ethernet (RJ45) jack on rear of drive
Complete Specifications & Product Details
Types of pump heads accepted0.1 to 650 RPM (up to 2 pump heads accepted)0.1 to 650 RPM Complete System with Easy-Load 77602-30 and C-Flex ULTRA® tubing 06434-73
Flow Range0.0006 to 19 LPM0.001 to 8.0 LPM
With EtherNet/IP and MasterflexLiveMFLX77421-00MFLX77966-00
With Profibus and MasterflexLiveMFLX77421-02MFLX77966-10
Drive Specifications
Speed control accuracy (resolution)±0.1% (0.1 rpm at 650 rpm)
Display7” capacitive TFT/RGB touch screen with in-plane switching (wide viewing angle); 800 x 480 pixels
External control - Input0 to 20 mA, 20 to 0 mA, 4 to 20 mA, 20 to 4 mA; 0 to 10V, 10 to 0 V, 0 to 5 V, 5 to 0 V; Start/Stop, Direction (CW/CCW), Prime via contact closure
External Control - Output0 to 20 mA, 20 to 0 mA, 4 to 20 mA, 20 to 4 mA; 0 to 10V, 10 to 0 V, 0 to 5 V, 5 to 0 V; Tach output (open collector), Motor running, Open head, Local/Remote, General alarm
MotorBLDC with encoder, continuous duty, reversible, 250 W (0.34 hp)
Open head sensorYes; extension cable included
Housing materials316 stainless steel housing and bezel with anodized aluminum base
Enclosure ratingIP66, NEMA 4X
Agency approvalsUL, cUL, CE
Warranty2 years
Dimensions (L x W x H)14" x 10.2" x 13.6" (35.6 x 26.0 x 34.6 cm)
Shipping weight19.5 kg (43 lb)
Power90 to 260 VAC, 50/60 Hz
I/P Pump Tubing Flow Ranges in LPM
rpmI/P Precision pump tubingI/P High-performance Precision pump tubing
I/P 26I/P 73I/P 82I/P 70I/P 88I/P 89
0.1 to 6500.0006 to 4.00.001 to 8.00.002 to 130.001 to 8.00.002 to 170.002 to 19
touchscreen-closeup.jpg
Set up is a breeze
Intuitive touch screen interface, even wearing gloves
  • Easy to set up and program
  • 7" capacitive TFT/RGB touch screen with a clear, wide viewing angle
  • Adjust parameters with a simple touch-even with gloves on
  • Operational modes include continuous run, volume dispense, and time dispense
  • Batch control and interval/off time in Volume and Time modes
  • Antidrip and speed ramping available in Volume mode-ensure accuracy and minimize sample loss
  • Easily save and recall up to 50 commonly used protocols, each with a unique name
  • Know when you hit your mark-graphical indicator lets you know when loaded tubing is optimally calibrated
Easy Integration and Scale-up
44030_1_closed-system-or-non-contaminating_100x100.png
Minimize risk with a closed system. Non-contaminating, low shear, and noninvasive makes this pump perfect for Biopharma and healthcare.
44030_2_Access-or-save-and-recall-programs.png
Three-tier access and the ability to save and recall programs make your fluidic process secure and accurate.
44030_3_scale-up.png
The I/P series is ideal for mimicking large-scale (up or out) systems, which allows quicker, consistent process optimization.
Avantor_Tubing_gradient-100.jpg
Precision manufactured pump tubing ensures accuracy, reliability, fluid compatibility and the regulatory approvals needed for critical process applications.
1.Continuous-Mode.png
Continuous Mode
2.Time-Mode.png
Time Mode
3.Volume-Dispense.png
Volume Dispense
4.Pressure-Sensor.png
Pressure Sensor
5.User-Access.png
User Access
6.Program-Save-and-Recall.png
Program Save & Recall
7.Network-Detection.png
Network Detection
mastersense-ls-analog-sm.png
Remote Mode
mastersense-ls-tubecalibration-sm.png
Tube Calibration
Take your process to the next level
MasterSense_I-P_MasterSense.png
  1. Perfect for sanitary environments-316 polished stainless steel housing IP66 NEMA 4X rated for washdown
  2. Integrated sensors monitor fluid pressure and alert before there is a potential for process failure
  3. Intuitive digital touchscreen with optimized menu navigation to store and recall standard protocols ensuring continuity - even with wet gloves on!
  4. Over-the-air firmware updates ensure optimization of your pump system without moving the system while MasterflexLive® cloud storage system securely stores, monitors and generates reports for all Masterflex pump systems
  5. Powerful performance with a broad flow range- 0.0006 to 26 LPM. Brushless, maintenance-free motor provides ±0.1% speed control accuracy with 6000:1 turndown ratio
    OTHER DRIVE OPTIONS
  6. Convenient open-head sensor and antidrip features. Accepts all I/P pump heads.
    SHOP I/P PUMP HEADS
  7. Graphical indicator validates optimal tube calibration. Choose from over 20 formulations and sizes of pump tubing.
    SHOP I/P PUMP TUBING
    DOWNLOAD FLYER