Orders LinkContactUsLinkComponent
water-testing-514459769-1920x768.jpg
Whatman™ filters for water testing
Water testing is essential for ensuring the safety and quality of water for human consumption and environmental and industrial applications, as well as for ensuring compliance with regulatory requirements. Accurate testing helps detect contaminants, measure chemical and biological components and maintain regulatory standards.
Whatman™ filters for water testing | Avantor
You can ensure that your environmental water testing meets the highest standards with Whatman™ filters, designed for precision and ease of use in a range of essential tests, including:
  • Solids analysis  
  • Metals analysis  
  • Dissolved ions testing
  • Dissolved organic carbon (DOC) content measurement
  • HPLC, UHPLC, ICP-MS  
  • Microbiological analysis  
Whatman™ filters are engineered to provide reliable results, even with challenging samples, to make each analysis quicker and more efficient. For example, Whatman’s 934-AH RTU glass fiber filters come prewashed and pre-weighed, saving significant time during suspended solids analysis.
For tough-to-filter samples, the GD/XP™ syringe filters, with an integrated dual-layer prefilter and a 0.45-µm membrane, streamline filtration while minimizing ion extractables, making them ideal for use in metal trace analysis by ICP-MS. Whether you're using Anotop™ IC, Puradisc™ Aqua 30, Spartan™, or Mini-UniPrep™, Whatman™ filters are designed to simplify your process and deliver precise results.
Whatman™ offers specific filters to guarantee reliable results and ease of use for analyses with critical samples. For example, the 934-AH RTU glass fiber filters are available prewashed and preweighed for considerable time savings when performing suspended solid analysis.
GD/XP™ syringe filters include an integrated prefiltration with dual-layer prefilter stack and one final 0.45-µm membrane to make the filtration of hard-to-filter samples easy. In addition, the pre-filter stack of polypropylene filters minimizes ion extractables, making these filters suitable for metal trace analysis by ICP-MS.
New: RTU (Ready-To-Use) family now includes filters
undefined
Product search tool
Avantor® offers a complete range of consumables, equipment and reagents for environmental analysis. Use our product search tool or contact your sales representative.
Request Whatman filter samples
Physical analysis 
You can measure total suspended, dissolved and volatile solids using glass fiber filters. The 934-AH RTU glass fiber filters are delivered ready for use.
Whatman-Filter-Paper-Grade-GFC-Filters_60533-540x390.jpg
Application: Filters for total suspended, dissolved and volatile solids analysis
  • Fine particle retention (1.2 µm)
Standard method compliance: EN 872
Filter name: GF/C
Filter type: Glass fiber filter
934-AH_13436-m-540x390.jpg
Application: Filters for total suspended solids analysis
  • High loading capacity
Standard method compliance: 2540D
Filter name: 934-AH
Filter type: Glass fiber filter
Grade-934-AH-RTU-Glass-Microfiber-Filter_13849-540x390.jpg
Application: Filters for total suspended, solids analysis
  • Ready for use: prewashed and preweighed
  • Packaged within an aluminum pan with a unique barcode
Standard method compliance: 2540D
Filter name: 934-AH RTU
Filter type: Glass fiber filter
Whatman-Filter-Paper-Grade-GFC-RTU-Microfiber-Glass-Filter-Binder-Free-Ready-to-Use_75628-540x390.jpg
Application: Filters for total suspended, dissolved and volatile solids analysis
  • Ready for use: prewashed, dried, cooled and weighed
  • Certified mass loss <0.017 mg/cm2 
Standard method compliance: EN 872
Filter name: GF/C RTU
Filter type: Glass fiber filter
Chemical analysis
Chemical analysis is commonly performed using analytical instrumentation. It is good practice to filter water prior to analysis to remove unwanted particles and protect delicate instrumentation from potentially damaging compounds.
Whatman™ offers filters for:
  • Dissolved heavy metals determination (GD/XP™ syringe filters, Polydisc™ GW in-line filters and Polycap™ GW capsule filters)
  • Dissolved ions testing (Anotop™ IC syringe filters)
  • Dissolved organic carbon (DOC) determination (Puradisc™ Aqua 30 syringe filters)
Whatman-GDXP-25-_58719-m-540x390.jpg
Application: Dissolved heavy metals analysis
  • For hard-to filter samples
  • Suitable for trace metals analysis
Filter name: GD/XP™
Filter type: Syringe filter
GE-Healthcare-1046340x-Polydisc-GW-Filter-540x390.jpg
Application: Dissolved heavy metals analysis
  • For groundwater samples
Filter name: Polydisc™ GW
Filter type: In-line filter
Polycap-GW-540x390.jpg
Application: Dissolved heavy metals analysis
  • For groundwater samples
Filter name: Polycap™ GW
Filter type: In-line filter
Whatman-Anotop-Syringe-Filters-Sterile_76005-540x390.jpg
Application: Dissolved ions testing
  • Certified and guaranteed low levels of anion leaching
Filter name: Anotop™ IC
Filter type: Syringe filter
GE-Healthcare-1046265x-Puradisc-Aqua-30-Syringe-Filter-540x390.jpg
Application: Dissolved organic carbon analysis
  • Prewashed
Filter name: Puradisc™ Aqua 30
Filter type: Syringe filters
HPLC/UHPLC and other analytical techniques
Use the following filters to protect your instruments and samples:
GE-Healthcare-10463xxx-SPARTAN-13-mm-HPLC-Certified-Syringe-Filter-540x390.jpg
Application: Low solids content
  • HPLC certified
Filter name: Spartan™
Filter type: Syringe filter
Whatman-Puradisc-Syringe-Filter-25mm-GFF-membrane-540x390.jpg
Application: Low solids content
  • Wide range of membranes, pore sizes and diameters
Filter name: Puradisc™
Filter type: Syringe filter
GE-Healthcare-GDX-13-Syringe-Filter-540x390.jpg
Application: Hard-to-filter samples
  • With a prefiltration glass filter stack 
  • Can hold three to seven times more sample than standard syringe filters
Filter name: GD/X™
Filter type: Syringe filter
Whatman-GDXP-25-_58719-540x390.jpg
Application: Hard-to-filter samples
  • With a polypropylene prefilter stack
  • Minimizes the levels of ion extractables
Filter name: GD/XP™
Filter type: Syringe filter
Mini-UniPrep-G2-Syringeless-Filter-540x390.jpg
Application: HPLC/GC autosamplers
  • All-in-one filter and autosampler vial
  • With glass autosampler vial
Filter name: Mini-UniPrep™ G2
Filter type: Syringeless filter
Syringeless-filters--Mini-UniPrep-Plastic-version-for-HPLCUHPLC-540x390.jpg
Application: HPLC/GC autosamplers
  • All-in-one filter and autosampler vial
  • With plastic autosampler vial
Filter name: Mini-UniPrep™
Filter type: Syringeless filter
Whatman-Puradisc-Syringe-Filter-25mm-GFF-membrane-540x390.jpg
Application: Spectrophotometry and chromatography
  • Wide range of membranes, pore sizes and diameters
Filter name: Puradisc™
Filter type: Glass fiber filter
Microbiological analysis
Microorganisms in water samples are collected using a microfiltration membrane filter. The membrane can be transferred onto a microbiological culture medium for further identification and/or quantification of microorganisms. The appropriate membrane filter choice will depend on the methodology being followed.
39092380-540x390.jpg
Application: Microbiological analysis
Filter name: MCE membrane
Filter type: MCE membrane filter
cytiva-microplus-540x390.jpg
Application: Microbiological analysis
  • High-flux cellulose nitrate
Filter name: MicroPlus
Filter type: NC membrane filter
Cytiva-new-image-907-540x390.jpg
Application: Microbiological analysis
  • For Legionella testing
Filter name: NL
Filter type: Nylon membrane filter
If you’re ready to move forward with your next innovation, Avantor® can help.