Orders LinkContactUsLinkComponent
PROVE-it.png
A major plus for routine analysis
New Spectroquant® Prove plus spectrophotometers with added speed, storage, and simplicity
As your trusted partner in analytical chemistry, we’re always working on the next innovation to enhance your analytical work. Our latest generation of spectrophotometers for your analysis does just that. The Spectroquant® Prove plus range is packed with features that make accuracy and precision simpler than ever.
Just PROVE it | Avantor
Positives of Prove plus for water, environmental and food & beverage analysis
  • Smoother functionality with an instantly responsive display and faster processing
  • Greater capacity to store more measurement results (up to 7000)
  • Extended warranty period of two years
Features and Benefits
Pre-programmed to work seamlessly with more than 150 Spectroquant® test kits, Spectroquant® Prove plus photometers are among the most trusted for your analysis.
  • Easy to use and pre-programmed with pre-defined reagents 
  • Reduced use of chemicals compared to traditional methods
  • Suitable for a broad measuring range per parameter (minimizes dilution work)
  • Pre-programmed for over 300 analysis methods
  • Cell size up to 10 cm with Prove 600 plus for high-sensitivity measurements
  • Instrument-supported AQA system for easy quality assurance
  • Automatic cell size recognition for cell & reagent tests
  • Live ID barcode for all cell & reagent tests (incl. lot and expiry data)
  • No adapter needed for the measurement of different cell sizes
  • Kits can be conveniently stored at room temperature
PROVE-it100a.png
Prove 100 plus
Routine VIS measurements in wastewater 
analysis and environmental testing 
Examples: Groundwater, effluents, soil
1.73026.0001
PROVE-it300.png
Prove 300 plus
Sensitive UV/VIS analysis of drinking water, 
food and beverages 
Examples: Drinking water, beverages, special beer applications, sugar, olive oil
1.73027.0001
PROVE-it600.png
Prove 600 plus
Complex UV/Vis measurements in process water testing
Examples: Power plants, cooler & boiler water
1.73028.0001