Orders LinkContactUsLinkComponent
DR300_Colorimeter_Backlight-955x590.jpg
Hach fast analysis
Hach offers a broad range of instrumentation and chemistries that have been carefully crafted for more than 80 years to make water analysis more accurate, faster, simpler and greener.
Hach fast analysis | Avantor
igX0kB7dbao-HD-540x390.jpg
Take 3 min to learn how your water analysis can:
  • Save time
  • Waste less
  • Have fewer mistakes
Play video
dulKCqFDJpw-HD-540x390.jpg
Take 4 min to learn how your water analysis can:
  • Save 75% more time
  • Waste less
  • Have fewer mistakes
Play video
SL1000 handheld parallel analyzer from Hach for streamlined water quality testing
Hach-SL1000-PPA-results_LPV443-200.jpg
Perform common water tests with fewer than half the manual steps when you use Chemkey® technology.
Provides 75% faster testing by using Chemkey® reagents instead of powder pillows or cuvette test methods.
Spectrophotometers and test kits from Hach
DR6000_spectrophotometer-200.jpg
Solutions that make daily routine water testing easier.
UV/visible spectrophotometers with radio frequency (RFID) technology to ensure high-speed wavelength scanning across the UV and visible spectrum.
Visible spectrophotometers ensure the reliability and traceability of results during routine analysis. Scans and tracks by using RFID tags.
DR1900 spectrophotometers are accurate and portable for testing drinking, waste and industrial waters.
Portable colorimeters from Hach
DR300_Colorimeter__Mobile-Connect-200.jpg
Effective and accurate handheld solutions:
  • DR300 pocket colorimeters for single-parameter testing 
  • The DR900 pocket colorimeter has 90 pre-programmed water testing methods
Simple, intuitive operation reduces potential manual errors to ensure accurate measurement data.
Intended for custom applications not included in standard analysis systems. Available in a range of wavelengths for custom calibrations.
The DR900 is a robust multi-parameter colorimeter configured for immediate use and specially designed for field applications.
Need more information about Hach fast analysis equipment?