Orders LinkContactUsLinkComponent
hichrom_sfc_600.jpg
Supercritical fluid chromatography
Avantor® supports supercritical fluid chromatography (SFC) with products from key suppliers to fully meet your specific analytical chromatography needs. SFC is a chromatographic technique that uses a supercritical fluid as the mobile phase. SFC has been increasingly adopted as an orthogonal technique to HPLC, particularly in the pharmaceutical industry. This is due in part to SFC's ability to combine high-throughput separation with a reduced amount of solvent. Additionally, SFC offers more rapid sample processing and drying down, lower costs and increased safety.
Supercritical fluid chromatography at Avantor® | Avantor
SFC has been commonly adopted for chiral and achiral chromatography and can be readily scaled to have fewer preparation steps than many HPLC processes.
Featured suppliers
Find out more about our range of chiral SFC products from Chiral Technologies.
Choose from a selection of Kromasil® SFC resources from Nouryon.
We offer a variety of achiral SFC columns from Princeton Chromatography.
Avantor® is here to help improve the efficiency and performance of your chromatography processes.
Learn more about how SFC could make your analytical lab more green.
Contact us