Orders LinkContactUsLinkComponent
hichrom_sec_600.jpg
Size exclusion chromatography at Avantor®
Avantor® offers size exclusion chromatography (SEC) columns to support the full range of chromatography techniques used by the analytical community.
Size exclusion chromatography at Avantor® | Avantor
SEC separates solution components by molecular size, with larger molecules eluting first. This technique can also be applied for characterizing the molecular weight distribution of polymers. Both silica- and polymer-based SEC columns are available.
Gel permeation chromatography (GPC) separates organic-soluble polymers/molecules using an organic solvent, while gel filtration chromatography (GFC) separates water-soluble polymers/molecules in aqueous solvents.
Featured suppliers
BioBasic™ SEC columns from Thermo Scientific are ideal for the high-efficiency gel filtration separation of proteins, peptides and other biological water-soluble molecules.
Learn more about the range of SEC and GPC products from Tosoh Bioscience, designed to meet the demanding needs of the pharma and biopharma industries.
Avantor® is ready to help improve the efficiency and performance of your chromatography processes.