Orders LinkContactUsLinkComponent
main page_sampaling_treaser_image.jpg
Sampling
An extensive range of high-quality sampling consumables and equipment to help you ensure a reliable analytical procedure.
Sampling | Avantor
Products
1stimage400x495.jpg
Samples must be contained in specific containers to avoid contamination. You will find here in wide choice a wide choice of sizes, shapes and specific uses to meet these very specific analyses.
2stimage400x495.jpg
Large choice of Polypropylene cylinders to perform on specific analysis.
Sampling-image400x495.jpg
These special tubes have been designed for easy filling and high performance sample preparation.
Sampling-image_400x495.jpg
Syringes for chromatography
These syringes are designed for the highest performance in applications such as liquid or gas chromatography and the handling of corrosive gases and liquids, radioactive materials, and sterile solutions. Syringes are gas-tight and leak-proof.
Sampling-image4400x495.jpg
Pasteur/Transfer pipettes
Completely control the small quantity of liquid being manually dispensed with the Pasteur and transfer pipettes. The all-in-one devices are inert to most fluids for maximum working flexibility. Squeezing the small or large bulb, releases homogeneous droplets from the transparent, narrow stems that may come graduated for precise biological spotting or filling applications. Due to the challenge of properly washing the bulb, Pasteur and transfer pipettes are available in disposable as well as reusable models.
Sampling-image6400x495.jpg
Balances and scales
High quality calibrated balances are simple to operate for quick weighing calculations. Using the latest technology available, scales provide reliable mass measurements with fast stabilization timing.