Orders LinkContactUsLinkComponent
hero blockimage EPA method 8325.jpg
EPA Method 8327 for Surface water, Wastewater or groundwater
Determination of selected PFAS in Surface water, Wastewater or groundwater by Liquid Chromatography/tandem mass spectrometry (LC-MS/MS).
EPA-Method-8327 | Avantor
Products
1stproductsamplepreparation.jpg
PFAS friendy vials
Specific polypropylene vials recommended when analysing polar compounds /analytes and dimensionally verified/inspected by automated optical and gauges to ensure critical dimensions and tolerances.
2stproductsamplepreparation.jpg
SPE for PFAS analysis
Special phase for SPE analysis. For enrichment of PFAS from water, textiles and sediments (contaminated soils). Polymeric combination phase with weak anion exchanger, highly porous, spherical particles.