Orders LinkContactUsLinkComponent
method 537.1 heromain page.jpg
EPA Method 537.1 for Drinking water
Determination of selected PFAS in Drinking water by solid phase extraction and Liquid Chromatography/tandem mass spectrometry (LC-MS/MS).
EPA Method 537.1 | Avantor
Products
2stproductsamplepreparation.jpg
PFAS friendy vials
Specific polypropylene vials recommended when analysing polar compounds /analytes and dimensionally verified/inspected by automated optical and gauges to ensure critical dimensions and tolerances.
2stproductsamplepreparation.jpg
SPE for PFAS analysis
Special phase for SPE analysis. For enrichment of PFAS from water, textiles and sediments (contaminated soils). Polymeric combination phase with weak anion exchanger, highly porous, spherical particles.
2stproductsamplepreparation.jpg
Caps for PFAS analysis
Special phase for SPE analysis. For enrichment of PFAS from water, textiles and sediments (contaminated soils). Polymeric combination phase with weak anion exchanger, highly porous, spherical particles.
2stproductsamplepreparation.jpg
LC Columns
Specific polypropylene vials recommended when analysing polar compounds /analytes and dimensionally verified/inspected by automated optical and gauges to ensure critical dimensions and tolerances.
2stproductsamplepreparation.jpg
GC Columns
Special phase for SPE analysis. For enrichment of PFAS from water, textiles and sediments (contaminated soils). Polymeric combination phase with weak anion exchanger, highly porous, spherical particles.