Orders LinkContactUsLinkComponent
method 1633 heromain page.jpg
EPA Method 1633 in aqueous, Solids, Biosolids and tissue samples by LC-MS/MS
Determination of selected PFAS in aqueous, Solids, Biosolids and tissue  samples by LC-MS/MS.
EPA Method 1633 | Avantor
Products
2stproductsamplepreparation.jpg
PFAS friendy vials
Specific polypropylene vials recommended when analysing polar compounds /analytes and dimensionally verified/inspected by automated optical and gauges to ensure critical dimensions and tolerances.
1stproductsamplepreparation.jpg
SPE for PFAS analysis
Special phase for SPE analysis. For enrichment of PFAS from water, textiles and sediments (contaminated soils). Polymeric combination phase with weak anion exchanger, highly porous, spherical particles.