Orders LinkContactUsLinkComponent
Bioprocessing Fermentation Process Workflow | Avantor
Bioprocessing Fermentation Process
fermentationworkflow.jpg
Masterflex supports the entire bioprocessing fluid path
Temperature Sensors
38728324-200x200.jpg
Single-Use Temperature Sensors
Sample Bottle
38732695-200.jpg
Tanks, vessels, and bottles store product between the stages of drug production. They are sterilized or involve a sterilized barrier between the vessel and the product (such as a tank liner) to mitigate the risk of contamination. Tank ports are pre-ordered to fit the needs of the application. Sample bottles will include a vent filter to avoid pressurizing the bottle during dispensation and cross-contamination.
View sample bottle assemblies
Vent Filter
Many biopharma applications require the use of vent filters on tank ventilation. Generally, these filters are sterilized to avoid risk of contamination. The filters also serve as a pressure stabilizer for the tank environment.
View vent filter products
Flow Sensors
masterflex-masterflex-ultrasonic-flow-sensors-6744.jpg
Masterflex Ultrasonic Flow Sensors
masterflex-masterflex-single-use-flow-tube-sensors-200.jpg
Masterflex® Single-Use Flow Tube Sensors
levitronix-leviflow-single-use-precision-ultrasoni.jpg
LEVIFLOW® Single-Use Precision Ultrasonic Flowmeters
levitronix-leviflow-clamp-on-ultrasonic-flow-senso.jpg
LEVIFLOW® Clamp-On Ultrasonic Flow Sensors
Pressure Sensors
38728332-200.jpg
PendoTECH® Single-Use Pressure Sensors
38728321-200.jpg
PendoTECH® Pressure Monitoring and Transmitting Systems