Orders LinkContactUsLinkComponent
safety-new-heroimage.png
Advanced Battery Science - Personal Protective Equipment & Safety
Avantor® offers a broad range of personal protective equipment to protect the operator when researching manufacturing or recycling.
Access our broad portfolio of products and solutions
Gloves
clove.jpg
A selection of gloves for hand protection from lab to production applications.
VWR® Nitrile
Ambidextrous gloves are soft and comfortable for everyday use. They protect hands from a broad range of chemicals. Beaded cuff allows easy donning.
Ideal for use in industrial and laboratory applications. Made of premium compounded nitrile for high manufacturing quality measures and product consistency.
Advanced nitrile material formulation features added thickness to provide excellent barrier protection. Gloves are produced in ISO certified facilities.
Three layer design for superior protection against harsh chemicals including acids, solvents and bases.
KC PURPLE NITRILE™ and PURPLE NITRILE-XTRA™
Nitrile gloves offer excellent tactile sensitivity and dexterity as well as superior fit and comfort.
Specially formulated with a nitrile compound that provides outstanding protection against a wide range of chemicals, including most solvents and acids.
Ideal for use in environments handling items or tools with a high risk of cuts, including steel or glass handling, assembly, maintenance and janitorial applications.
Polyurethane coating provides basic cut protection and grip. Ideal for handling small components, or situations that require dexterity in a dry and/or dirty environment.
Liners provide protection and comfort when used with latex, nitrile and PVC gloves. Fully launderable for reusability and economy.
Neoprene-coated gloves combine the flexibility and chemical resistance of neoprene with the comfort and heat resistance of a soft thermal liner.
Experience EnerGel™ infused gloves, designed to hydrate skin and protect against long-term glove usage, with biodegradable EcoTek® technology.
Eye and Face Protection
Safety-glasses.jpg
Safety glasses and goggles, visitor spectacles and face shields for every situation.
Wrap-around sport styling, designed for men and women features eight base spherical lenses providing excellent side protection.
VWR® Sleek Safety Glasses
Lightweight and sleekly designed safety glasses. Meets ANSI Z87.1 standards.
Inner temple coated with elastomer for excellent grip and soft cushioning with impact-resistance.
Economical and compact, designed to fit over prescription glasses. Meets ANSI Z87.1 standards.
VWR® Ladies Pink Safety Glasses
Stylish protective eyewear combining fashion, fit and function. Most suitable for small facial contours and provides 99.9% UV protection.
Super lightweight wraparound eyewear for men and women combines a sleek appearance with rugged durability.
Stylish and secure protection with an array of comfort features to enable extended use.
Sporty, frameless, lightweight design enhances comfort. No-brow design increases upward and peripheral vision.
Glasses Uvex Seismic offers a range of diopter strengths for workers needing increased magnification for reading, inspection and other fine detail work.
Strong, lightweight, polycarbonate lenses with Duramass® scratch-resistant coating filter out 99% of UV radiation.
Fashion and function combine with sleek styling that offers a lightweight, comfortable fit.
Wide vision goggles provide high levels of comfort and maximum splash protection.
Economical non sterile safety goggles are designed to protect from hazards with a flexible ridge around a PVC frame.
Designed to increase comfort when users are required to wear other personal protective equipment.
A soft, pliable frame conforms to the face for comfort and is easy to clean. Indirect ventilation. Can be worn over most prescription eyeglasses.
Provides full face protection from debris and splash hazards. Ideal for use in many environments ranging from the lab to industrial applications.
Perfect for use in environments with light particle, splash, or spray, the VWR® Disposable Face Shield is a cost effective and reliable way to keep covered.
Honeywell Safety Uvex Bionic® Face Shields
Lightweight face shields feature a clear polycarbonate visor that provides enhanced protection from airborne debris and increased visibility.
Respiratory Protection
Respiratory-protection.jpg
A selected choice of disposable respirators, half and full face masks, Powered Air Purifying Respirators (PAPR).
VWR® Disposable N95 Particulate Respirators
Disposable Respirators combine excellent safety and comfort features to ensure reliable protection and a lightweight, secure fit.
Up to 31% more breathable than the leading competitive style and up to 75% more breathable than the NIOSH minimum requirement.
FLUIDSHIELD* N95 filter respirators are NIOSH approved and are designed to provide an effective facial fit as required for proper respiratory protection.
Economical respirator provides reliable worker protection against certain solid and liquid aerosols that do not contain oil.
3M™ 7500 Series Reusable Half Mask Respirators
These durable, reusable respirators allow the user to change the filter to protect against particulates, gases and vapors.
Certain Organic Vapors/Acid Gas/P100 approved for use against chlorine, hydrogen chloride, sulfur dioxide, hydrogen fluoride, hydrogen sulfide (escape only).
3M™ 6000 Series Full Facepiece Respirators with accessories
Reusable, lightweight full face pieces; respirators feature easy-to-adjust head straps for maximum comfort.
All-in-one system ready for use, including one HEPA filter. Loose-fitting face piece designed for use with a stethoscope.
Up to 8cfm, 33% greater airflow within the hood and suit. The hood attaches to protective clothing with clips, for a secure fit.
Head and Hearing Protection
Head_Phone.jpg
Safety helmets, earplugs and earmuffs.
Featuring a four-point suspension, these hats are lightweight, low-profile and have a low center of gravity, providing exceptional balance.
CEN10™ Safety Helmets are designed to provide wearers with the reliability, comfort and toughness expected from a Bullard industrial hard hat.
An exclusive AdB coating on the ear pods not only increases attenuation, but also repels dirt and allows the pod to be washed and reused.
Pod Plug™ design eliminates the fear of hand-to-ear contamination by removing the need to touch or roll down the foam during insertion.
Pre-shaped foam earplugs feature a smooth outer skin for maximum user comfort. Meet ANSI S3.19 standard. CE marked. CSA certified.
Dielectric three-position, ultra-lightweight earmuffs are ideal for prolonged use and comfort.
Unique dual-cup design and soft, foam-filled cushions provide noise reduction and comfort.
Disposable Apparel
Disposable.jpg
Disposable apparels suited for basic up to the most demanding working conditions to protect against chemicals, particles, aersols and flames.
VWR® Basic SPP Beard Covers
Manufactured from a spun-bonded polypropylene (SPP) fabric that is strong, lightweight and breathable.
Ideally suited for medical, industrial, laboratory, electronic and pharmaceutical environments.
VWR® Basic SMS Coveralls
Garments feature inset sleeves, front zipper, tapered collar and elastic cuffs, ankles and back.
VWR® Signature™ Hooded Coveralls made with DuPont™ Tyvek® IsoClean® Material
A unique flash spinning process creates an excellent barrier against dry particulates, nonhazardous liquids and microorganisms.
DuPont™ Tyvek® 400 Coveralls with Respirator Hood and Attached Skid Resistant Boots
Composed of flash spun high density polyethylene which creates a unique, nonwoven material available only from DuPont.
VWR® Maximum Fluid-Impervious Butterfly-Style Shoe Covers
Shoe covers are rigorously tested and manufactured in an ISO Certified facility to ensure each garment meets quality standards and performs to specification.
DuPont™ Tychem® 4000 Bib Aprons
DuPont™ Tychem® 4000 delivers effective protection against a range of chemical environments.
Cost-efficient, disposable over-garment designed to help protect and preserve primary flame resistant garments.
VWR® Maximum Lab Coats
Manufactured from a specially formulated, breathable microporous fabric for exceptional comfort and protection.
VWR® Basic Industrial Lab Coats
A lightweight, single-use protective option available in a variety of colors and sizes.
Reusable Apparel and Workwear
Reusable.jpg
Lab coats, ESD coats and jackets, High visibility garments, flame resistant coveralls to wear and protect in any situation.
VWR® Unisex Technology Lab Coat, Reusable
Unisex Technology lab coats are designed for use in research environments, quality-control laboratories and industrial areas.
Manufactured from breathable and comfortable 98% polyester fabric and 2% carbon fabric. Provides anti-static protection.
ESD jackets feature a snap front closure, wrist-adjustment snaps, three roomy patch pockets and lapel style collars
High-visibility, long-sleeved T-Shirts are compliant with ANSI ISEA 107/2010 Class 2 standards.
High-visibility, short-sleeve polo shirts are designed to help keep wearers seen and safe.
Footwear
Shoe.jpg
Cleanroom clogs, Shoes and boots to work confidently in any environment.
Designed for comfort and durability in cleanroom environments. Suitable for autoclave sterilization.
100% waterproof knee boots made from high performance PVC compound upper and a PVC/Nitrile sole.