Orders LinkContactUsLinkComponent
Sterilization-Wraps | Avantor
Sterilization Wraps
bhdsheet2-removebg.png
Designed for steam sterilization; but also compatible with ETO or gamma irradiation
SMS wrap.png
Strong, rip and tear resistant 50 gsm SMS material
SMS wrap.png
Strong, rip and tear resistant 88 gsm SMS material
onestep300-1.png
These 2-ply, 50 gsm wrappers provide an excellent microbial barrier. Ideal for pharmaceutical production applications.
Manufactured and packaged in a certified ISO 8 Cleanroom.
onestep600-1.png
These 2-ply, 88 gsm wrappers provide an excellent microbial barrier. Ideal for pharmaceutical production applications.
Manufactured and packaged in a certified ISO 8 Cleanroom.
079.png
Drapeable 1422A Tyvek® Wrappers are an improvement over cellulose or SMS based wrapping materials for steam sterilization.
079.png
This new, uncoated Tyvek® eliminates the potential of residue deposits on equipment, post sterilization.