Tork Xpress® H2 Compressed Soft Multifold Hand Towels, White
Catálogo # SCAA100889
Proveedor: Essity
New Product
Especificaciones
- Para:Compatible with Tork H2 dispensers
- Presentación:12 packs, 225 paper hand towels each; 2 ply
- Color:blanco
- Descripción:Premium multifold hand towels, refill
- l×a:80×212 mm (Folded size); 240×212 mm (Unfolded size)
Especificaciones
Acerca de este artículo
Tork Xpress® compressed soft multifold hand towels white H2, premium, 2-ply, Z-fold. Bring extra comfort to guests in high-end establishments with these soft and thick hand towels.
Reduce paper towel consumption in washrooms and improve hygiene with one-at-a-time dispensing
- Reduce waste and increase cost savings - these hand towels are quick-drying and absorbent, so guests don't use as many
-
Maximise the space - use these compressed paper hand towels with the compact Tork Xpress® dispenser in low-to-medium traffic washrooms
Premium quality, compatible with H2 dispensers
Save time restocking dispensers with 50% compressed paper hand towels that double dispenser capacity. Guests enjoy a premium experience with these soft and absorbent hand towels. One-at-a-time dispensing helps improve hygiene and reduce waste as guests only touch the towel they take. Our compact dispensers fit into small spaces for maximum convenience.
Certificaciones:
FSC MIX Credit, SA-COC-008266; EU Ecolabel, SE/004/001.