Dishwashing Detergent, ProCare Universal 81 C
Catálogo # MIEL12456130
Proveedor: MIELE
New Product
Especificaciones
- Descripción:Machine descaler, ProCare Universal 81 C
- Env:250 g
- Rango de temperatura:5...40 °C
Especificaciones
Acerca de este artículo
Machine descaler, acidic, for removing persistent limescale deposits.
- Removal of silicate deposits and other acid-soluble impurities
- Prolongs service life of heater elements
- Very gentle on materials
- Very good material compatibility
- Specially developed for Miele Professional machines
Contains citric acid and does not contain phosphorus compounds.
Certificaciones: GHS/CLP compliant.