Print-on Hook Labels BradyGrip® with Velcro® Hook for BradyJet™ J7300 Printer
BRDY176590
: Brady
New Product
- Tipo:B-2414
- Color:blanco
- Dimensiones:25,40W mm × 22,86L m
- Material:Poliéster
- Modelo:JC-1000-2414
- Env:1
BradyGrip® Print-on Hook labels combine the power of on-demand colour labelling and the flexibility of VELCRO® material for unmatched identification of wires and cables.
- Keep wires and cables clearly identified to breeze through installations, troubleshooting, and repairs
- Use colour to add strong visual cues when locating wire and cable bundles
- Removable and repositionable
- Inkjet receptive polyester material
- Designed for indoor use
Easily apply, reposition and remove this exclusive print-and-place material created from a Brady and VELCRO® brand partnership. Simply print a unique label, then apply to a VELCRO® Brand ONE-WRAP® roll. Built-in LabelSense™ technology enables cartridge and printer communication for fast changeovers, automatic label setup, and seamless software interaction.
For use with the BradyJet™ J7300 colour label printer. Ink cartridges are required to print this material; use part numbers P-CMY-1 and P-K-1. VELCRO® Brand ONE-WRAP® product not included with purchase.