Entrada de pedido
Spain
Orders LinkContactUsLinkComponent
Mouse HRG ELISA Kit

Mouse HRG ELISA Kit

Mouse HRG ELISA kit, 96 Tests
Catalog # ANTIA79438-96
Supplier:   Antibodies.com
Mouse HRG ELISA Kit
Catalog # ANTIA79438-96
Supplier:  Antibodies.com
CAS Number:  
Mouse HRG ELISA Kit
Mouse HRG ELISA Kit

Mouse HRG ELISA kit, 96 Tests

CAS Number:  
Price
625,00 €
Log in for availability
1 * 96 Tests
Product Details & Documents

Mouse HRG ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse HRG in serum, plasma, tissue homogenates, and other biological fluids.

  • Ready To Use ELISA Kit
  • Detection Range: 0,156 to 10 ng/ml
  • Sensitivity: 0,094 ng/ml
  • Sample Type: Serum, plasma, tissue homogenates, and other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Mouse HRG ELISA Kit (A79438) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse HRG in serum, plasma, tissue homogenates, and other biological fluids. An antibody specific for HRG has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the HRG present in each sample is bound to the wells by the immobilized antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-HRG Antibody, which binds the captured HRG present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Streptavidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualize the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of HRG captured in each well. The concentration of HRG can then be calculated by reading the O.D. absorbance at 450nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, lyophilised standard, sample dilution buffer, biotinylated detection antibody (100X), antibody dilution buffer, HRP-Streptavidin conjugate (SABC) (100X), SABC dilution buffer, TMB substrate, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.

Specifications

Specifications

  • Env:
    96 Tests
  • Descripción:
    Mouse HRG ELISA kit
  • Tipo de la muestra:
    Serum, plasma, tissue homogenates and other biological fluids
  • Primary antibody reactivity:
    Mouse
  • Reactividad cruzada:
    Mouse HRG ELISA Kit exhibits high specificity and excellent specificity for the detection of mouse HRG. No significant cross-reactivity or interference between HRG and analogues was observed.
  • Duración del ensayo:
    Multiple steps
  • Rango de detección:
    0,156 - 10 ng/ml
  • Tipo de ensayo:
    Sandwich
  • Shelf life:
    Store for 6 months at 4 °C
  • Principio de ensayo:
    Quantitative
  • Formato:
    Pre-coated
  • Tiempo para resultados:
    4 h 30 min
  • Volumen de la muestra:
    100 μl
  • Cat. No.:
    ANTIA79438-96
  • Temperatura de almacenamiento:
    4 °C
  • Objetivo:
    HRG
  • Sensibilidad:
    0,094 ng/ml
  • Regulatory status:
    RUO
  • Método de detección:
    Colorimetric

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.