JOE Dye qPCR Calibration Plate *Optimised for ABI7500 Fast 96-Well*
Catálogo # AATB67012
Proveedor: AAT Bioquest
New Product
Especificaciones
- Color del marco:blanco
- Nº de pocillos:96
- Perfil:Low
- Faldón:Semi-skirted
- Color del pocillo:blanco
- Descripción:JOE Dye qPCR calibration plate
- Con código de barras:Si
- Env:1
Especificaciones
Acerca de este artículo
The JOE dye qPCR calibration plate can be used to maintain the 7500 real-time PCR system with fast 96-well block.
- The qPCR calibration plate might significantly improve qPCR results with multiplexing by more accurately representing fluorescent spectra used in your real-time experiments
- This calibration plate is ready to use without any additional preparation steps required
For most qPCR instruments, the necessary calibrations should be run at least every six months.
Precaución: Avoid freeze-thawing plates. It is not recommended to use the plates more than 3 times. Plates can be stored for up to 12 months. Freeze (<−15 °C), and minimise light exposure.