TAMRA Dye qPCR Calibration Plate *Optimised for ABI7500 Fast 96-Well*
Catalog # AATB67000
Supplier: AAT BIOQUEST
New Product
Specifications
- Color del marco:blanco
- Nº de pocillos:96
- Perfil:Low
- Faldón:Semi-skirted
- Color del pocillo:blanco
- Descripción:TAMRA Dye qPCR calibration plate
- Con código de barras:Si
- Env:1
Specifications
About this item
The TAMRA dye qPCR calibration plate can be used to calibrate and maintain the 7500 real-time PCR system with fast 96-well block.
- The qPCR calibration plate might significantly improve qPCR results with multiplexing by more accurately representing fluorescent spectra used in your real-time experiments
- This calibration plate is ready to use without any additional preparation steps required
For most qPCR instruments, the necessary calibrations should be run at least every six months.
Caution: Avoid freeze-thawing plates. It is not recommended to use the plates more than 3 times. Plates can be stored for up to 12 months. Freeze (<−15 °C), and minimise light exposure.